Gene Transcripts

Gene Symbol
FRAS1
Entrez Gene ID
80144
Gene Name
Fraser extracellular matrix complex subunit 1
Gene Aliases
Chromosome Location
Chr. 4: 78057323 - 78544269 on Build GRCh38
UniGene ID
Hs.369448
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025074.6 NP_079350.5 45 6849
XM_006714316.2 XP_006714379.1 45 7096
GenBank mRNA AB040933.2 - 7 1094

View Full Product Details