Gene Transcripts

Gene Symbol
TEKT1
Entrez Gene ID
83659
Gene Name
tektin 1
Gene Aliases
Chromosome Location
Chr. 17: 6799981 - 6831741 on Build GRCh38
UniGene ID
Hs.462108
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053285.1 NP_444515.1 6 852
XM_011524027.2 XP_011522329.1 6 848
GenBank mRNA AF357879.1 - 6 801
AK292818.1 - 6 817
AK304345.1 - 5 651
BC014599.2 - 6 852 4824860

View Full Product Details