Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_032119.3 | NP_115495.3 | 31 | 6858 | |
NR_003149.1 | 31 | 6858 | |||
XM_017009963.1 | XP_016865452.1 | 31 | 6977 | ||
XM_017009964.1 | XP_016865453.1 | 31 | 6974 | ||
XM_017009965.1 | XP_016865454.1 | 31 | 6859 | ||
XM_017009966.1 | XP_016865455.1 | 30 | 6896 | ||
XM_017009967.1 | XP_016865456.1 | 30 | 6766 | ||
XM_017009968.1 | XP_016865457.1 | 31 | 6977 | ||
XM_017009969.1 | XP_016865458.1 | 31 | 6977 | ||
XM_017009970.1 | XP_016865459.1 | 31 | 6977 | ||
XM_017009971.1 | XP_016865460.1 | 31 | 6977 | ||
XM_017009972.1 | XP_016865461.1 | 3 | 342 | ||
XM_017009973.1 | XP_016865462.1 | 8 | 1574 | ||
XM_017009974.1 | XP_016865463.1 | 31 | 6977 | ||
GenBank mRNA | AF435925.1 | - | 31 | 6858 | |
CD633594.1 | - | 3 | 379 |