Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001206998.1 | NP_001193927.1 | 2 | 378 | |
NM_032173.3 | NP_115549.2 | 2 | 171 | ||
XM_011530435.2 | XP_011528737.1 | 3 | 367 | ||
XM_011530436.2 | XP_011528738.1 | 4 | 422 | ||
XM_011530437.1 | XP_011528739.1 | 3 | 1092 | ||
XM_011530438.2 | XP_011528740.1 | 3 | 608 | ||
XM_017028990.1 | XP_016884479.1 | 3 | 863 | ||
GenBank mRNA | AB051436.1 | - | 2 | 243 | |
AK096397.1 | - | 2 | 171 | ||
CR456397.1 | - | 2 | 151 |