Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001229.4 | NP_001220.2 | 9 | 1447 | |
NM_001278054.1 | NP_001264983.1 | 5 | 997 | ||
NM_032996.3 | NP_127463.2 | 9 | 1278 | ||
NR_102732.1 | 9 | 1505 | |||
NR_102733.1 | 9 | 1307 | |||
XM_005246014.2 | XP_005246071.1 | 9 | 1131 | ||
GenBank mRNA | AB015653.1 | - | 5 | 773 | |
AB020979.1 | - | 5 | 752 | ||
AB209147.1 | - | 8 | 2303 | ||
AF093130.1 | - | 5 | 752 | ||
AF110376.1 | - | 6 | 752 | ||
AJ781267.1 | - | 9 | 1062 | ||
AK292080.1 | - | 9 | 1219 | ||
AK292111.1 | - | 9 | 1223 | ||
AK303743.1 | - | 9 | 1214 | ||
AY732490.2 | - | 9 | 1260 | ||
BC002452.2 | - | 9 | 1208 | 3343958 | |
BC006463.1 | - | 9 | 1297 | 3542075 | |
BT006911.1 | - | 9 | 1202 | ||
U56390.1 | - | 9 | 1205 | ||
U60521.1 | - | 9 | 1247 |