Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_032427.3 | NP_115803.1 | 5 | 4143 | |
XM_011543023.2 | XP_011541325.1 | 5 | 5978 | ||
XM_011543024.2 | XP_011541326.1 | 5 | 2683 | ||
GenBank mRNA | AB058722.1 | - | 5 | 4134 | |
AK308157.1 | - | 6 | 3909 | ||
AY040322.1 | - | 5 | 4134 | ||
BC143529.1 | - | 5 | 2884 | 9052038 | |
BC152449.1 | - | 5 | 4134 | 8862580 | |
CR627398.1 | - | 1 | 3139 |