Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001289158.1 | NP_001276087.1 | 6 | 645 | |
NM_001289159.1 | NP_001276088.1 | 4 | 423 | ||
NM_032571.4 | NP_115960.2 | 7 | 801 | ||
XM_011528374.2 | XP_011526676.1 | 6 | 692 | ||
XM_017027383.1 | XP_016882872.1 | 2 | 95 | ||
GenBank mRNA | AF239764.1 | - | 7 | 738 | |
AK290909.1 | - | 7 | 725 | ||
AK298653.1 | - | 4 | 349 | ||
AL832663.1 | - | 7 | 802 | ||
AY358817.1 | - | 6 | 542 | ||
BC121130.2 | - | 7 | 723 | ||
BC121131.2 | - | 7 | 723 | ||
BC128053.1 | - | 7 | 723 | ||
BC148282.2 | - | 7 | 723 |