Gene Transcripts
- Gene Symbol
- WDR73
- Entrez Gene ID
- 84942
- Gene Name
- WD repeat domain 73
- Gene Aliases
- GAMOS, HSPC264
- Chromosome Location
- Chr. 15: 84640682 - 84654343 on Build GRCh38
- UniGene ID
- Hs.745027
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_032856.3 |
NP_116245.2
|
1, 2 |
111 |
|
NR_130945.1 |
|
1, 2 |
111 |
|
NR_130947.1 |
|
1, 2 |
111 |
|
GenBank mRNA |
AF161382.1 |
- |
1, 2 |
42 |
|
AK027794.1 |
- |
1, 2 |
48 |
|
AK055729.1 |
- |
1, 2 |
48 |
|
AK225068.1 |
- |
1, 2 |
51 |
|
AK225199.1 |
- |
1, 2 |
48 |
|
AK295372.1 |
- |
1, 2 |
42 |
|
BC063392.1 |
- |
1, 2 |
79 |
5168550 |
BX446522.2 |
- |
1, 2 |
41 |
|
CD514627.1 |
- |
1, 2 |
34 |
|
View Full Product Details