Gene Transcripts

Gene Symbol
STK19
Entrez Gene ID
8859
Gene Name
serine/threonine kinase 19
Gene Aliases
D6S60, D6S60E, G11, HLA-RP1, RP1
Chromosome Location
Chr. 6: 31971175 - 31981446 on Build GRCh38
UniGene ID
Hs.654371
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_004197.1 NP_004188.1 6 1013
NM_032454.1 NP_115830.1 6 1025
NR_026717.1 6 1198
GenBank mRNA BC110347.1 - 6 1198
BC121128.1 - 5 1032 40120738
BC121129.1 - 5 1031 40120740
BC143833.1 - 5 1068 9052348
BC143835.1 - 5 1080
L26260.1 - 6 1013
X77386.1 - 5 619

View Full Product Details