Gene Transcripts

Gene Symbol
ACTN3
Entrez Gene ID
89
Gene Name
actinin alpha 3 (gene/pseudogene)
Gene Aliases
Chromosome Location
Chr. 11: 66546395 - 66563329 on Build GRCh38
UniGene ID
Hs.654432
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001104.3 NP_001095.2 17 2175
NM_001258371.2 NP_001245300.2 17 2323
NR_047663.2 17 2175
GenBank mRNA AK125851.1 - 17 2709
AK313556.1 - 17 2113
BC096323.2 - 11 1235
BC099647.3 - 17 2096 40008220
BC099648.1 - 7 844
BC099649.3 - 17 2096 40008223
CX787471.1 - 2 169
M86407.1 - 17 2096

View Full Product Details