Gene Transcripts
- Gene Symbol
- KNSTRN
- Entrez Gene ID
- 90417
- Gene Name
- kinetochore localized astrin/SPAG5 binding protein
- Gene Aliases
- C15orf23, HSD11, SKAP, TRAF4AF1
- Chromosome Location
- Chr. 15: 40382721 - 40394288 on Build GRCh38
- UniGene ID
- Hs.525796
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001142762.1 |
NP_001136234.1
|
7, 8 |
874 |
|
NM_033286.3 |
NP_150628.3
|
8, 9 |
936 |
|
GenBank mRNA |
AK027408.1 |
- |
8, 9 |
695 |
|
AK125765.1 |
- |
8, 9 |
893 |
|
AK298156.1 |
- |
7, 8 |
600 |
|
AK301887.1 |
- |
8, 9 |
936 |
|
AY652615.1 |
- |
8, 9 |
835 |
|
BC004543.2 |
- |
8, 9 |
645 |
|
BC014060.2 |
- |
8, 9 |
645 |
|
BC045739.1 |
- |
8, 9 |
717 |
|
BC064344.1 |
- |
8, 9 |
644 |
|
BC107802.1 |
- |
8, 9 |
702 |
|
BC118559.1 |
- |
8, 9 |
865 |
40023827 |
BC118639.1 |
- |
8, 9 |
865 |
40023825 |
U81002.1 |
- |
8, 9 |
692 |
|
View Full Product Details