Gene Transcripts

Gene Symbol
LRSAM1
Entrez Gene ID
90678
Gene Name
leucine rich repeat and sterile alpha motif containing 1
Gene Aliases
CMT2P, RIFLE, TAL
Chromosome Location
Chr. 9: 127451486 - 127503501 on Build GRCh38
UniGene ID
Hs.495188
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005373.3 NP_001005373.1 17 1579
NM_001005374.3 NP_001005374.1 16 1284
NM_001190723.2 NP_001177652.1 17 1579
NM_138361.5 NP_612370.3 16 1838
XM_006717316.3 XP_006717379.1 17 1579
XM_017015283.1 XP_016870772.1 17 2578
XM_017015284.1 XP_016870773.1 17 1574
GenBank mRNA AK056203.1 - 17 1561
AK056305.1 - 5 424
AK226106.1 - 17 1565
AY358830.1 - 17 1544
BC009239.2 - 16 1284 2988664

View Full Product Details