Gene Transcripts
- Gene Symbol
- LRSAM1
- Entrez Gene ID
- 90678
- Gene Name
- leucine rich repeat and sterile alpha motif containing 1
- Gene Aliases
- CMT2P, RIFLE, TAL
- Chromosome Location
- Chr. 9: 127451486 - 127503501 on Build GRCh38
- UniGene ID
- Hs.495188
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001005373.3 |
NP_001005373.1
|
22, 23 |
2061 |
|
NM_001005374.3 |
NP_001005374.1
|
21, 22 |
1766 |
|
NM_001190723.2 |
NP_001177652.1
|
21, 22 |
1980 |
|
NM_138361.5 |
NP_612370.3
|
21, 22 |
2320 |
|
XM_017015283.1 |
XP_016870772.1
|
22, 23 |
3060 |
|
XM_017015284.1 |
XP_016870773.1
|
22, 23 |
2056 |
|
GenBank mRNA |
AK056203.1 |
- |
21, 22 |
1962 |
|
AK056305.1 |
- |
10, 11 |
911 |
|
AK091589.1 |
- |
21, 22 |
2068 |
|
AK097094.1 |
- |
2, 3 |
1521 |
|
AK226106.1 |
- |
22, 23 |
2047 |
|
AY358830.1 |
- |
22, 23 |
2026 |
|
BC009239.2 |
- |
21, 22 |
1766 |
2988664 |
View Full Product Details