Gene Transcripts

Gene Symbol
ZMYM5
Entrez Gene ID
9205
Gene Name
zinc finger MYM-type containing 5
Gene Aliases
HSPC050, MYM, ZNF198L1, ZNF237
Chromosome Location
Chr. 13: 19823482 - 19863636 on Build GRCh38
UniGene ID
Hs.530988
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039649.2 NP_001034738.1 3 343
NM_001039650.2 NP_001034739.1 3 343
NM_001142684.1 NP_001136156.1 3 343
XM_005266593.3 XP_005266650.1 3 730
XM_005266594.2 XP_005266651.1 2 158
XM_006719894.2 XP_006719957.1 3 453
XM_011535309.1 XP_011533611.1 3 702
XM_011535310.1 XP_011533612.1 3 702
XM_017020849.1 XP_016876338.1 2 265
XM_017020850.1 XP_016876339.1 3 332
XM_017020851.1 XP_016876340.1 3 702
GenBank mRNA AJ133352.1 - 3 231
AJ133353.1 - 3 231
AJ133354.1 - 3 231
AJ133355.1 - 3 231
AK312724.1 - 3 175
BC007048.1 - 3 215 3681047
BI561630.1 - 3 350
BT006687.1 - 1 78

View Full Product Details