Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_033518.3 | NP_277053.2 | 13 | 1097 | |
XM_005272694.3 | XP_005272751.2 | 12 | 1087 | ||
XM_005272695.4 | XP_005272752.3 | 12 | 1089 | ||
XM_005272697.2 | XP_005272754.2 | 12 | 1040 | ||
XM_005272698.4 | XP_005272755.2 | 12 | 978 | ||
XM_006724569.3 | XP_006724632.1 | 13 | 1097 | ||
XM_017029960.1 | XP_016885449.1 | 12 | 1009 | ||
XM_017029961.1 | XP_016885450.1 | 12 | 981 | ||
GenBank mRNA | AF276889.1 | - | 12 | 1762 | |
AF318358.1 | - | 9 | 1208 | ||
AK094788.1 | - | 12 | 1016 | ||
BC019246.2 | - | 12 | 947 | 3506044 | |
BC027721.2 | - | 8 | 638 |