Gene Transcripts

Gene Symbol
MS4A3
Entrez Gene ID
932
Gene Name
membrane spanning 4-domains A3
Gene Aliases
CD20L, HTM4
Chromosome Location
Chr. 11: 60056628 - 60071116 on Build GRCh38
UniGene ID
Hs.99960
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001031666.1 NP_001026836.1 4, 5 440
NM_001031809.1 NP_001026979.1 5, 6 611
NM_006138.4 NP_006129.4 6, 7 749
XM_011545363.2 XP_011543665.1 5, 6 496
GenBank mRNA AY095480.1 - 4, 5 483
BC008487.1 - 6, 7 694 4102067
BE564041.1 - 4, 5 461
BF210971.1 - 482
L35848.1 - 6, 7 719

View Full Product Details