Gene Transcripts
- Gene Symbol
- ZRANB2
- Entrez Gene ID
- 9406
- Gene Name
- zinc finger RANBP2-type containing 2
- Gene Aliases
- ZIS, ZIS1, ZIS2, ZNF265
- Chromosome Location
- Chr. 1: 71063291 - 71081289 on Build GRCh38
- UniGene ID
- Hs.194718
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_005455.4 |
NP_005446.2
|
2, 3 |
391 |
|
NM_203350.2 |
NP_976225.1
|
2, 3 |
391 |
|
GenBank mRNA |
AB209568.1 |
- |
2, 3 |
104 |
|
AF065391.1 |
- |
2, 3 |
131 |
|
AF065392.1 |
- |
2, 3 |
131 |
|
AK222858.1 |
- |
2, 3 |
116 |
|
AL136945.1 |
- |
2, 3 |
113 |
|
AU100296.1 |
- |
2, 3 |
165 |
|
BC033648.1 |
- |
2, 3 |
137 |
|
BC039814.1 |
- |
2, 3 |
137 |
6062647 |
DA523601.1 |
- |
2, 3 |
391 |
|
View Full Product Details