Gene Transcripts

Gene Symbol
QKI
Entrez Gene ID
9444
Gene Name
QKI, KH domain containing RNA binding
Gene Aliases
Hqk, QK, QK1, QK3, hqkI
Chromosome Location
Chr. 6: 163414486 - 163578596 on Build GRCh38
UniGene ID
Hs.510324
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001301085.1 NP_001288014.1 6 1369
NM_006775.2 NP_006766.1 6 1393
NM_206853.2 NP_996735.1 6 1393
NM_206854.2 NP_996736.1 6 1393
NM_206855.2 NP_996737.1 6 1393
XM_011536258.2 XP_011534560.1 6 897
XM_011536259.2 XP_011534561.1 6 876
XM_011536260.2 XP_011534562.1 5 1291
XM_017011502.1 XP_016866991.1 6 900
XM_017011503.1 XP_016866992.1 6 897
XM_017011504.1 XP_016866993.1 6 876
GenBank mRNA AB067798.1 - 6 1394
AB067799.1 - 6 1394
AB067800.1 - 6 1394
AB067801.1 - 6 1394
AF142422.1 - 6 953
AF272349.1 - 6 1275
AK027309.1 - 6 800
AK295235.1 - 3 404
AK308784.1 - 3 341
AL050218.1 - 1 86
AY780788.1 - 6 806
BC012222.1 - 5 700
BC019917.2 - 6 881 3921310
DQ323998.1 - 6 818

View Full Product Details