Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_004890.2 | NP_004881.2 | 6 | 550 | |
XM_006721600.2 | XP_006721663.1 | 6 | 1175 | ||
GenBank mRNA | AF047437.1 | - | 6 | 527 | |
AK123065.1 | - | 1329 | |||
AK312174.1 | - | 6 | 526 | ||
BC011934.2 | - | 6 | 519 | 4330461 | |
BP308951.1 | - | 6 | 549 |