Gene Transcripts

Gene Symbol
NOS1AP
Entrez Gene ID
9722
Gene Name
nitric oxide synthase 1 adaptor protein
Gene Aliases
6330408P19Rik, CAPON
Chromosome Location
Chr. 1: 162069791 - 162370023 on Build GRCh38
UniGene ID
Hs.731942
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001126060.1 NP_001119532.2 2 2838
NM_001164757.1 NP_001158229.1 10 1728
NM_014697.2 NP_055512.1 10 1743
GenBank mRNA AB007933.2 - 10 1743
AF037070.1 - 2 297
AY841899.1 - 2 2838
BC041382.2 - 10 1745
BC112295.1 - 10 1444 8327763
BC143771.1 - 10 1429 9052285
BC143780.1 - 11 1542

View Full Product Details