Gene Symbol | Transcript Accession | Exon Location | Assay Location | Protein ID |
---|---|---|---|---|
SLC12A6 | NM_001042494.1 | 26 | 5751 | NP_001035959.1 |
NM_001042495.1 | 26 | 5426 | NP_001035960.1 | |
NM_001042496.1 | 26 | 5817 | NP_001035961.1 | |
NM_001042497.1 | 24 | 5520 | NP_001035962.1 | |
NM_005135.2 | 25 | 5390 | NP_005126.1 | |
NM_133647.1 | 25 | 5565 | NP_598408.1 | |
XM_006720793.3 | 24 | 6300 | XP_006720856.1 | |
AK056019.1 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1052620 | Chr15:32612402 - 34538731 on Build GRCh38 | Gain | LOC100131315 ARHGAP11A LOC101928134 GREM1 MIR1233-1 LOC105370757 TMCO5B NOP10 PGBD4 NUTM1 RYR3 LOC101928042 EMC4 GOLGA8A SLC12A6 CHRM5 EMC7 FMN1 SCG5 AVEN MIR1233-2 GOLGA8B LPCAT4 KATNBL1 |
esv33337 | Chr15:31689863 - 34575097 on Build GRCh38 | Gain+Loss | GOLGA8K ARHGAP11A LOC101928134 LOC105370940 LOC105370757 WHAMMP1 NUTM1 RYR3 CHRNA7 GOLGA8A LOC100996255 EMC7 SCG5 LOC100653133 AVEN MIR1233-2 LPCAT4 KATNBL1 LOC100131315 GREM1 MIR1233-1 TMCO5B ULK4P1 LOC101060588 GOLGA8N NOP10 PGBD4 LOC101928042 EMC4 SLC12A6 CHRM5 GOLGA8O FMN1 GOLGA8B |
esv3569431 | Chr15:33564570 - 35080307 on Build GRCh38 | Gain | LOC101928174 MIR1233-1 GJD2 NOP10 PGBD4 NUTM1 RYR3 EMC4 GOLGA8A SLC12A6 CHRM5 EMC7 AVEN ZNF770 MIR1233-2 GOLGA8B LPCAT4 ACTC1 AQR KATNBL1 |
dgv4456n54 | Chr15:32604149 - 34557061 on Build GRCh38 | Gain | ARHGAP11A LOC101928134 LOC105370757 NUTM1 RYR3 GOLGA8A EMC7 SCG5 AVEN MIR1233-2 LPCAT4 KATNBL1 LOC100131315 GREM1 MIR1233-1 TMCO5B GOLGA8N NOP10 PGBD4 LOC101928042 EMC4 SLC12A6 CHRM5 FMN1 GOLGA8B |