Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 113205441-113205938 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NCAM1
Entrez Gene ID
4684
Gene Name
neural cell adhesion molecule 1
Gene Aliases
CD56, MSK39, NCAM
UniGene
Hs.503878
Gene Chromosome Location
Chr.11: 112961247-113278436 on Build GRCh38
Transcripts
NM_000615.6 NM_001076682.3 NM_001242607.1 NM_001242608.1 NM_181351.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6130943 c.460G>T p.V154F Chr.11: 113205666-113205666 on Build GRCh38
4856358 c.386G>A p.R129Q Chr.11: 113205565-113205565 on Build GRCh38
4856356 c.359G>A p.R120Q Chr.11: 113205565-113205565 on Build GRCh38
5418691 c.487+10G>C p.? Chr.11: 113205676-113205676 on Build GRCh38
6722600 c.318G>T p.Q106H Chr.11: 113205524-113205524 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1006494776 Chr.11: 113205856-113205856 on Build GRCh38
rs550834882 Chr.11: 113205775-113205775 on Build GRCh38
rs782000032 Chr.11: 113205774-113205774 on Build GRCh38
rs782167976 Chr.11: 113205544-113205544 on Build GRCh38
rs782623986 Chr.11: 113205573-113205573 on Build GRCh38
rs959335343 Chr.11: 113205531-113205531 on Build GRCh38
rs182204242 Chr.11: 113205911-113205911 on Build GRCh38
rs1025519582 Chr.11: 113205891-113205891 on Build GRCh38
rs367901170 Chr.11: 113205565-113205565 on Build GRCh38
rs531203109 Chr.11: 113205857-113205857 on Build GRCh38
rs782333827 Chr.11: 113205510-113205510 on Build GRCh38
rs782323888 Chr.11: 113205643-113205643 on Build GRCh38
rs782177193 Chr.11: 113205623-113205623 on Build GRCh38
rs371317616 Chr.11: 113205474-113205474 on Build GRCh38
rs560214539 Chr.11: 113205673-113205673 on Build GRCh38
rs781976487 Chr.11: 113205505-113205505 on Build GRCh38
rs781924056 Chr.11: 113205686-113205686 on Build GRCh38
rs782158279 Chr.11: 113205674-113205674 on Build GRCh38
rs568469218 Chr.11: 113205749-113205749 on Build GRCh38
rs782788205 Chr.11: 113205538-113205538 on Build GRCh38
rs782230366 Chr.11: 113205581-113205581 on Build GRCh38
rs929677262 Chr.11: 113205795-113205795 on Build GRCh38
rs781810668 Chr.11: 113205551-113205551 on Build GRCh38
rs375478322 Chr.11: 113205641-113205641 on Build GRCh38
rs782676092 Chr.11: 113205609-113205609 on Build GRCh38
rs200921858 Chr.11: 113205582-113205582 on Build GRCh38
rs369715213 Chr.11: 113205648-113205648 on Build GRCh38
rs782342694 Chr.11: 113205483-113205483 on Build GRCh38
rs782421120 Chr.11: 113205678-113205678 on Build GRCh38
rs957963041 Chr.11: 113205717-113205717 on Build GRCh38
rs1013583443 Chr.11: 113205874-113205874 on Build GRCh38
rs142086348 Chr.11: 113205739-113205739 on Build GRCh38
rs782124790 Chr.11: 113205514-113205514 on Build GRCh38
rs782071211 Chr.11: 113205700-113205700 on Build GRCh38
rs202238052 Chr.11: 113205820-113205820 on Build GRCh38
rs910913241 Chr.11: 113205732-113205732 on Build GRCh38
rs1041227886 Chr.11: 113205741-113205741 on Build GRCh38
rs551301385 Chr.11: 113205863-113205863 on Build GRCh38
rs781902410 Chr.11: 113205488-113205488 on Build GRCh38
rs887914114 Chr.11: 113205830-113205830 on Build GRCh38
rs888040830 Chr.11: 113205467-113205467 on Build GRCh38
rs377357078 Chr.11: 113205652-113205652 on Build GRCh38
rs191287596 Chr.11: 113205542-113205542 on Build GRCh38
rs985320985 Chr.11: 113205720-113205720 on Build GRCh38

View Full Product Details