Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 52446561-52447064 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KRT6B
Entrez Gene ID
3854
Gene Name
keratin 6B
Gene Aliases
CK-6B, CK6B, K6B, KRTL1, PC2, PC4
UniGene
Hs.708950
Gene Chromosome Location
Chr.12: 52446651-52452126 on Build GRCh38
Transcripts
NM_005555.3

SNP Detail

SNP ID Chromosome Location
rs553864968 Chr.12: 52446963-52446963 on Build GRCh38
rs1035599636 Chr.12: 52446787-52446787 on Build GRCh38
rs921535517 Chr.12: 52446684-52446684 on Build GRCh38
rs535888045 Chr.12: 52447003-52447003 on Build GRCh38
rs530008221 Chr.12: 52446825-52446825 on Build GRCh38
rs973202585 Chr.12: 52447019-52447019 on Build GRCh38
rs776834961 Chr.12: 52446600-52446600 on Build GRCh38
rs540139576 Chr.12: 52446840-52446840 on Build GRCh38
rs921630217 Chr.12: 52446666-52446666 on Build GRCh38
rs907800771 Chr.12: 52446739-52446739 on Build GRCh38
rs944257612 Chr.12: 52446679-52446679 on Build GRCh38
rs974516340 Chr.12: 52446920-52446920 on Build GRCh38
rs116436520 Chr.12: 52446771-52446771 on Build GRCh38
rs569009342 Chr.12: 52446965-52446965 on Build GRCh38
rs544756853 Chr.12: 52446978-52446978 on Build GRCh38
rs987816101 Chr.12: 52446727-52446727 on Build GRCh38
rs921515234 Chr.12: 52446977-52446977 on Build GRCh38
rs561715124 Chr.12: 52446761-52446761 on Build GRCh38
rs778963143 Chr.12: 52446764-52446764 on Build GRCh38
rs906552218 Chr.12: 52446822-52446822 on Build GRCh38
rs924748233 Chr.12: 52446937-52446937 on Build GRCh38
rs977181847 Chr.12: 52446894-52446894 on Build GRCh38
rs532425818 Chr.12: 52446939-52446939 on Build GRCh38
rs1031641288 Chr.12: 52446850-52446850 on Build GRCh38
rs552132387 Chr.12: 52446958-52446958 on Build GRCh38
rs772264432 Chr.12: 52446957-52446957 on Build GRCh38
rs1003565476 Chr.12: 52446838-52446838 on Build GRCh38
rs559974200 Chr.12: 52446855-52446855 on Build GRCh38
rs1048413800 Chr.12: 52446709-52446709 on Build GRCh38
rs892725244 Chr.12: 52446852-52446852 on Build GRCh38
rs560980582 Chr.12: 52446773-52446773 on Build GRCh38
rs745722883 Chr.12: 52446744-52446744 on Build GRCh38
rs553678436 Chr.12: 52446656-52446656 on Build GRCh38
rs1021302780 Chr.12: 52446859-52446859 on Build GRCh38
rs536958694 Chr.12: 52446630-52446630 on Build GRCh38
rs545233358 Chr.12: 52446738-52446738 on Build GRCh38
rs1048350395 Chr.12: 52447026-52447026 on Build GRCh38
rs909900755 Chr.12: 52447032-52447032 on Build GRCh38
rs924780901 Chr.12: 52446669-52446669 on Build GRCh38
rs748270391 Chr.12: 52446872-52446872 on Build GRCh38
rs1045031461 Chr.12: 52446807-52446807 on Build GRCh38
rs73308613 Chr.12: 52446767-52446767 on Build GRCh38
rs897683277 Chr.12: 52446768-52446768 on Build GRCh38
rs781067320 Chr.12: 52446702-52446702 on Build GRCh38
rs1041316220 Chr.12: 52446681-52446681 on Build GRCh38
rs187031962 Chr.12: 52446663-52446663 on Build GRCh38

View Full Product Details