Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 10637470-10637964 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CASZ1
Entrez Gene ID
54897
Gene Name
castor zinc finger 1
Gene Aliases
CAS11, CST, SRG, ZNF693, dJ734G22.1
UniGene
Hs.439894
Gene Chromosome Location
Chr.1: 10636602-10796676 on Build GRCh38
Transcripts
NM_001079843.2 XM_017001539.1 XM_017001540.1 XM_017001541.1

SNP Detail

SNP ID Chromosome Location
rs1008216022 Chr.1: 10637716-10637716 on Build GRCh38
rs538649088 Chr.1: 10637868-10637868 on Build GRCh38
rs142845529 Chr.1: 10637785-10637785 on Build GRCh38
rs12404480 Chr.1: 10637562-10637562 on Build GRCh38
rs746379625 Chr.1: 10637495-10637495 on Build GRCh38
rs537056136 Chr.1: 10637684-10637684 on Build GRCh38
rs761746653 Chr.1: 10637647-10637647 on Build GRCh38
rs984495331 Chr.1: 10637797-10637800 on Build GRCh38
rs752778925 Chr.1: 10637851-10637851 on Build GRCh38
rs558972562 Chr.1: 10637699-10637699 on Build GRCh38
rs199867187 Chr.1: 10637822-10637822 on Build GRCh38
rs985266381 Chr.1: 10637778-10637778 on Build GRCh38
rs765052797 Chr.1: 10637744-10637744 on Build GRCh38
rs867290124 Chr.1: 10637927-10637927 on Build GRCh38
rs906636442 Chr.1: 10637862-10637862 on Build GRCh38
rs908863589 Chr.1: 10637808-10637808 on Build GRCh38
rs567321963 Chr.1: 10637858-10637858 on Build GRCh38
rs1031519674 Chr.1: 10637677-10637677 on Build GRCh38
rs1056491776 Chr.1: 10637636-10637636 on Build GRCh38
rs965854695 Chr.1: 10637500-10637500 on Build GRCh38
rs775079096 Chr.1: 10637511-10637511 on Build GRCh38
rs1022748582 Chr.1: 10637668-10637668 on Build GRCh38
rs563284604 Chr.1: 10637646-10637646 on Build GRCh38
rs567186092 Chr.1: 10637823-10637823 on Build GRCh38
rs530400127 Chr.1: 10637592-10637592 on Build GRCh38
rs995886633 Chr.1: 10637624-10637624 on Build GRCh38
rs1009602888 Chr.1: 10637935-10637935 on Build GRCh38
rs1037216988 Chr.1: 10637817-10637817 on Build GRCh38
rs763866012 Chr.1: 10637620-10637620 on Build GRCh38
rs144310260 Chr.1: 10637807-10637807 on Build GRCh38
rs541717997 Chr.1: 10637861-10637861 on Build GRCh38
rs183063689 Chr.1: 10637503-10637503 on Build GRCh38
rs918819567 Chr.1: 10637828-10637828 on Build GRCh38
rs879231006 Chr.1: 10637822-10637823 on Build GRCh38
rs191609779 Chr.1: 10637915-10637915 on Build GRCh38
rs1017796996 Chr.1: 10637725-10637725 on Build GRCh38
rs1009529372 Chr.1: 10637680-10637680 on Build GRCh38
rs914281172 Chr.1: 10637523-10637523 on Build GRCh38
rs188185377 Chr.1: 10637622-10637622 on Build GRCh38
rs1027005276 Chr.1: 10637627-10637627 on Build GRCh38
rs1044685524 Chr.1: 10637850-10637850 on Build GRCh38
rs182954626 Chr.1: 10637939-10637939 on Build GRCh38
rs750398418 Chr.1: 10637767-10637767 on Build GRCh38
rs981245715 Chr.1: 10637676-10637676 on Build GRCh38
rs971244707 Chr.1: 10637673-10637673 on Build GRCh38
rs956134924 Chr.1: 10637899-10637899 on Build GRCh38
rs931605209 Chr.1: 10637829-10637829 on Build GRCh38
rs1050333922 Chr.1: 10637560-10637560 on Build GRCh38
rs1002635914 Chr.1: 10637863-10637863 on Build GRCh38
rs139937925 Chr.1: 10637714-10637714 on Build GRCh38
rs534482520 Chr.1: 10637784-10637784 on Build GRCh38
rs533664463 Chr.1: 10637559-10637559 on Build GRCh38
rs760192800 Chr.1: 10637532-10637532 on Build GRCh38
rs924279093 Chr.1: 10637496-10637496 on Build GRCh38
rs1012364110 Chr.1: 10637651-10637651 on Build GRCh38
rs551436928 Chr.1: 10637635-10637635 on Build GRCh38
rs976221757 Chr.1: 10637760-10637760 on Build GRCh38
rs878987907 Chr.1: 10637607-10637607 on Build GRCh38
rs566301546 Chr.1: 10637575-10637575 on Build GRCh38
rs948694056 Chr.1: 10637823-10637823 on Build GRCh38
rs891333834 Chr.1: 10637574-10637574 on Build GRCh38
rs966630712 Chr.1: 10637731-10637731 on Build GRCh38
rs990201870 Chr.1: 10637708-10637708 on Build GRCh38
rs1048863291 Chr.1: 10637843-10637843 on Build GRCh38

View Full Product Details