Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 10639794-10640288 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CASZ1
Entrez Gene ID
54897
Gene Name
castor zinc finger 1
Gene Aliases
CAS11, CST, SRG, ZNF693, dJ734G22.1
UniGene
Hs.439894
Gene Chromosome Location
Chr.1: 10636602-10796676 on Build GRCh38
Transcripts
NM_001079843.2 XM_005263479.3 XM_017001539.1 XM_017001540.1 XM_017001541.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5842942 c.4412G>A p.R1471H Chr.1: 10639810-10639810 on Build GRCh38
4618942 c.4256C>T p.A1419V Chr.1: 10639966-10639966 on Build GRCh38
4020083 c.4246C>T p.R1416W Chr.1: 10639976-10639976 on Build GRCh38
1199687 c.4394G>A p.G1465D Chr.1: 10639828-10639828 on Build GRCh38
5626522 c.4404C>T p.F1468F Chr.1: 10639818-10639818 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754061163 Chr.1: 10639908-10639908 on Build GRCh38
rs867701505 Chr.1: 10640197-10640197 on Build GRCh38
rs375864672 Chr.1: 10639944-10639944 on Build GRCh38
rs775684437 Chr.1: 10640047-10640047 on Build GRCh38
rs370505461 Chr.1: 10639970-10639970 on Build GRCh38
rs750789845 Chr.1: 10639998-10639998 on Build GRCh38
rs1010268702 Chr.1: 10640144-10640144 on Build GRCh38
rs745537977 Chr.1: 10639957-10639957 on Build GRCh38
rs369777674 Chr.1: 10639813-10639813 on Build GRCh38
rs377572273 Chr.1: 10639884-10639884 on Build GRCh38
rs1028084012 Chr.1: 10640131-10640131 on Build GRCh38
rs910613011 Chr.1: 10640230-10640230 on Build GRCh38
rs772002077 Chr.1: 10640018-10640018 on Build GRCh38
rs773077535 Chr.1: 10640173-10640173 on Build GRCh38
rs904963944 Chr.1: 10640049-10640049 on Build GRCh38
rs200311968 Chr.1: 10640032-10640032 on Build GRCh38
rs778313706 Chr.1: 10640080-10640080 on Build GRCh38
rs1035205138 Chr.1: 10639814-10639814 on Build GRCh38
rs750879806 Chr.1: 10640064-10640064 on Build GRCh38
rs367680843 Chr.1: 10639986-10639986 on Build GRCh38
rs757568860 Chr.1: 10639945-10639945 on Build GRCh38
rs759781062 Chr.1: 10639977-10639977 on Build GRCh38
rs373399036 Chr.1: 10639858-10639858 on Build GRCh38
rs781563409 Chr.1: 10639950-10639950 on Build GRCh38
rs777674412 Chr.1: 10640095-10640095 on Build GRCh38
rs761502789 Chr.1: 10639886-10639886 on Build GRCh38
rs773030997 Chr.1: 10640023-10640023 on Build GRCh38
rs772938935 Chr.1: 10639976-10639976 on Build GRCh38
rs747304331 Chr.1: 10639840-10639840 on Build GRCh38
rs756544841 Chr.1: 10640068-10640068 on Build GRCh38
rs778230683 Chr.1: 10639836-10639836 on Build GRCh38
rs957790143 Chr.1: 10640115-10640115 on Build GRCh38
rs770665100 Chr.1: 10639842-10639842 on Build GRCh38
rs768450888 Chr.1: 10639880-10639880 on Build GRCh38
rs766786573 Chr.1: 10640071-10640071 on Build GRCh38
rs746784633 Chr.1: 10640024-10640024 on Build GRCh38
rs974415624 Chr.1: 10640204-10640204 on Build GRCh38
rs1050714209 Chr.1: 10639882-10639882 on Build GRCh38
rs775596134 Chr.1: 10639980-10639980 on Build GRCh38
rs968414490 Chr.1: 10640174-10640174 on Build GRCh38
rs564447983 Chr.1: 10640073-10640073 on Build GRCh38
rs766371464 Chr.1: 10640260-10640260 on Build GRCh38
rs181557754 Chr.1: 10639987-10639987 on Build GRCh38
rs545744774 Chr.1: 10639827-10639827 on Build GRCh38
rs558627995 Chr.1: 10640045-10640045 on Build GRCh38
rs986499605 Chr.1: 10640160-10640160 on Build GRCh38
rs778426739 Chr.1: 10640016-10640016 on Build GRCh38
rs771774588 Chr.1: 10639975-10639975 on Build GRCh38
rs766688041 Chr.1: 10639887-10639887 on Build GRCh38
rs746210310 Chr.1: 10639966-10639966 on Build GRCh38
rs544322406 Chr.1: 10639911-10639911 on Build GRCh38
rs774154742 Chr.1: 10639968-10639968 on Build GRCh38
rs768118953 Chr.1: 10640051-10640051 on Build GRCh38
rs988141274 Chr.1: 10639828-10639828 on Build GRCh38
rs201472163 Chr.1: 10639868-10639868 on Build GRCh38
rs374524422 Chr.1: 10640003-10640003 on Build GRCh38
rs757735053 Chr.1: 10639990-10639990 on Build GRCh38
rs775684219 Chr.1: 10640026-10640026 on Build GRCh38
rs529350884 Chr.1: 10640164-10640164 on Build GRCh38
rs748054379 Chr.1: 10640017-10640017 on Build GRCh38
rs769532690 Chr.1: 10639962-10639962 on Build GRCh38
rs779755173 Chr.1: 10639965-10639965 on Build GRCh38
rs968154002 Chr.1: 10639869-10639869 on Build GRCh38
rs754448546 Chr.1: 10640009-10640009 on Build GRCh38
rs752733658 Chr.1: 10639918-10639918 on Build GRCh38
rs749506491 Chr.1: 10640105-10640105 on Build GRCh38
rs966137860 Chr.1: 10640253-10640253 on Build GRCh38
rs754607166 Chr.1: 10640076-10640076 on Build GRCh38
rs528434079 Chr.1: 10640092-10640092 on Build GRCh38
rs769444743 Chr.1: 10639865-10639865 on Build GRCh38
rs142747673 Chr.1: 10640036-10640036 on Build GRCh38
rs753212200 Chr.1: 10639815-10639815 on Build GRCh38
rs1024256187 Chr.1: 10640069-10640069 on Build GRCh38
rs910644284 Chr.1: 10639890-10639890 on Build GRCh38
rs1040915264 Chr.1: 10639958-10639958 on Build GRCh38
rs762925857 Chr.1: 10639985-10639985 on Build GRCh38
rs758942977 Chr.1: 10639937-10639937 on Build GRCh38
rs371896402 Chr.1: 10640097-10640097 on Build GRCh38
rs955068152 Chr.1: 10640153-10640153 on Build GRCh38
rs762588032 Chr.1: 10640229-10640229 on Build GRCh38
rs763019646 Chr.1: 10639875-10639875 on Build GRCh38
rs770057294 Chr.1: 10640025-10640025 on Build GRCh38
rs201451822 Chr.1: 10640111-10640111 on Build GRCh38
rs373591122 Chr.1: 10639978-10639978 on Build GRCh38
rs370732393 Chr.1: 10639885-10639885 on Build GRCh38
rs561774145 Chr.1: 10640119-10640119 on Build GRCh38
rs747659122 Chr.1: 10640085-10640085 on Build GRCh38
rs768024968 Chr.1: 10639991-10639991 on Build GRCh38
rs780692802 Chr.1: 10639843-10639843 on Build GRCh38
rs768892130 Chr.1: 10640106-10640106 on Build GRCh38
rs555615473 Chr.1: 10640132-10640132 on Build GRCh38
rs375316064 Chr.1: 10639910-10639910 on Build GRCh38
rs976565047 Chr.1: 10640268-10640268 on Build GRCh38
rs140215862 Chr.1: 10640031-10640031 on Build GRCh38
rs780918239 Chr.1: 10640103-10640103 on Build GRCh38
rs755883738 Chr.1: 10640000-10640000 on Build GRCh38

View Full Product Details