Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 11960560-11961061 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PLOD1
Entrez Gene ID
5351
Gene Name
procollagen-lysine,2-oxoglutarate 5-dioxygenase 1
Gene Aliases
EDS6, LH, LH1, LLH, PLOD
UniGene
Hs.75093
Gene Chromosome Location
Chr.1: 11934667-11975542 on Build GRCh38
Transcripts
NM_000302.3 NM_001316320.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4941716 c.1013C>T p.A338V Chr.1: 11960683-11960683 on Build GRCh38
6359039 c.1023C>T p.G341G Chr.1: 11960693-11960693 on Build GRCh38
4941717 c.1154C>T p.A385V Chr.1: 11960683-11960683 on Build GRCh38
6506927 c.1206G>A p.R402R Chr.1: 11960735-11960735 on Build GRCh38
1294810 c.1057G>A p.E353K Chr.1: 11960727-11960727 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs986115260 Chr.1: 11960927-11960927 on Build GRCh38
rs776480771 Chr.1: 11960725-11960725 on Build GRCh38
rs562510011 Chr.1: 11960777-11960777 on Build GRCh38
rs565820157 Chr.1: 11960947-11960947 on Build GRCh38
rs374036329 Chr.1: 11960741-11960741 on Build GRCh38
rs763906693 Chr.1: 11960740-11960740 on Build GRCh38
rs758298439 Chr.1: 11960766-11960766 on Build GRCh38
rs769508543 Chr.1: 11960787-11960788 on Build GRCh38
rs202022200 Chr.1: 11960784-11960784 on Build GRCh38
rs770695057 Chr.1: 11960719-11960719 on Build GRCh38
rs368754769 Chr.1: 11960608-11960608 on Build GRCh38
rs780999460 Chr.1: 11960806-11960806 on Build GRCh38
rs952206910 Chr.1: 11961021-11961021 on Build GRCh38
rs745814613 Chr.1: 11960807-11960807 on Build GRCh38
rs752755933 Chr.1: 11960600-11960601 on Build GRCh38
rs757083515 Chr.1: 11960796-11960796 on Build GRCh38
rs200166434 Chr.1: 11960701-11960701 on Build GRCh38
rs769217332 Chr.1: 11960732-11960732 on Build GRCh38
rs944966464 Chr.1: 11960656-11960656 on Build GRCh38
rs748787201 Chr.1: 11960810-11960810 on Build GRCh38
rs1000820922 Chr.1: 11960668-11960668 on Build GRCh38
rs779712341 Chr.1: 11960790-11960791 on Build GRCh38
rs777716638 Chr.1: 11960768-11960770 on Build GRCh38
rs562856531 Chr.1: 11960617-11960617 on Build GRCh38
rs766824430 Chr.1: 11960760-11960760 on Build GRCh38
rs34707621 Chr.1: 11960606-11960607 on Build GRCh38
rs767051420 Chr.1: 11960663-11960663 on Build GRCh38
rs1032781250 Chr.1: 11960765-11960765 on Build GRCh38
rs754351882 Chr.1: 11960667-11960667 on Build GRCh38
rs775007891 Chr.1: 11960733-11960733 on Build GRCh38
rs746868590 Chr.1: 11960707-11960707 on Build GRCh38
rs140159343 Chr.1: 11960696-11960696 on Build GRCh38
rs915784395 Chr.1: 11960962-11960962 on Build GRCh38
rs1022351597 Chr.1: 11960902-11960902 on Build GRCh38
rs892868306 Chr.1: 11961025-11961025 on Build GRCh38
rs769707466 Chr.1: 11960808-11960808 on Build GRCh38
rs1011165837 Chr.1: 11960867-11960867 on Build GRCh38
rs769966142 Chr.1: 11960899-11960899 on Build GRCh38
rs377080927 Chr.1: 11960767-11960767 on Build GRCh38
rs369599447 Chr.1: 11960780-11960780 on Build GRCh38
rs892334267 Chr.1: 11960854-11960861 on Build GRCh38
rs949973819 Chr.1: 11960965-11960965 on Build GRCh38
rs761174430 Chr.1: 11960744-11960744 on Build GRCh38
rs768008712 Chr.1: 11960641-11960641 on Build GRCh38
rs551839180 Chr.1: 11960639-11960639 on Build GRCh38
rs746959665 Chr.1: 11960613-11960613 on Build GRCh38
rs199908943 Chr.1: 11960596-11960596 on Build GRCh38
rs560631473 Chr.1: 11961037-11961037 on Build GRCh38
rs757044881 Chr.1: 11960786-11960787 on Build GRCh38
rs777545904 Chr.1: 11960705-11960705 on Build GRCh38
rs142329815 Chr.1: 11960630-11960630 on Build GRCh38
rs972621978 Chr.1: 11960689-11960689 on Build GRCh38
rs1019327718 Chr.1: 11960881-11960881 on Build GRCh38
rs150627428 Chr.1: 11960648-11960648 on Build GRCh38
rs199733836 Chr.1: 11961013-11961014 on Build GRCh38
rs548817976 Chr.1: 11961024-11961024 on Build GRCh38
rs751358120 Chr.1: 11960772-11960773 on Build GRCh38
rs547939219 Chr.1: 11960922-11960922 on Build GRCh38
rs772939391 Chr.1: 11960609-11960609 on Build GRCh38
rs111398100 Chr.1: 11960769-11960769 on Build GRCh38
rs972533556 Chr.1: 11960909-11960909 on Build GRCh38
rs202051470 Chr.1: 11960605-11960605 on Build GRCh38
rs768771562 Chr.1: 11960598-11960598 on Build GRCh38
rs755056846 Chr.1: 11960693-11960693 on Build GRCh38
rs747209300 Chr.1: 11960795-11960795 on Build GRCh38
rs987772931 Chr.1: 11960820-11960820 on Build GRCh38
rs934357281 Chr.1: 11960990-11960990 on Build GRCh38
rs147536755 Chr.1: 11960655-11960655 on Build GRCh38
rs748344499 Chr.1: 11960697-11960697 on Build GRCh38
rs59578134 Chr.1: 11960987-11960987 on Build GRCh38
rs778023285 Chr.1: 11960793-11960793 on Build GRCh38
rs35656581 Chr.1: 11960729-11960729 on Build GRCh38
rs370305686 Chr.1: 11960734-11960734 on Build GRCh38
rs188848701 Chr.1: 11960951-11960951 on Build GRCh38
rs367592901 Chr.1: 11960766-11960766 on Build GRCh38
rs1018977687 Chr.1: 11960955-11960955 on Build GRCh38
rs991321417 Chr.1: 11960960-11960960 on Build GRCh38
rs112904654 Chr.1: 11960986-11960986 on Build GRCh38
rs376257665 Chr.1: 11960981-11960981 on Build GRCh38
rs200825919 Chr.1: 11960640-11960640 on Build GRCh38
rs748374204 Chr.1: 11960607-11960607 on Build GRCh38
rs1054557835 Chr.1: 11961007-11961007 on Build GRCh38
rs759025771 Chr.1: 11960618-11960618 on Build GRCh38

View Full Product Details