Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 50909911-50910416 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RNASEH2B
Entrez Gene ID
79621
Gene Name
ribonuclease H2 subunit B
Gene Aliases
AGS2, DLEU8
UniGene
Hs.306291
Gene Chromosome Location
Chr.13: 50909678-50970462 on Build GRCh38
Transcripts
NM_001142279.2 NM_024570.3 XM_011535229.1 XM_011535230.2 XM_011535231.2 XM_011535233.2 XM_017020747.1 NR_046552.1

Gene Symbol
RNASEH2B-AS1
Entrez Gene ID
100874255
Gene Name
RNASEH2B antisense RNA 1
Gene Aliases
-
UniGene
Hs.741501
Gene Chromosome Location
Chr.13: 50882378-50910712 on Build GRCh38
Transcripts
NM_001142279.2 NM_024570.3 XM_011535229.1 XM_011535230.2 XM_011535231.2 XM_011535233.2 XM_017020747.1 NR_046552.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6363600 c.19T>A p.C7S Chr.13: 50910095-50910095 on Build GRCh38
6310477 c.41G>A p.R14Q Chr.13: 50910117-50910117 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs111392805 Chr.13: 50910205-50910205 on Build GRCh38
rs1047859776 Chr.13: 50910070-50910070 on Build GRCh38
rs913827642 Chr.13: 50909966-50909966 on Build GRCh38
rs542198163 Chr.13: 50910270-50910270 on Build GRCh38
rs966576861 Chr.13: 50910005-50910005 on Build GRCh38
rs919933802 Chr.13: 50909993-50909993 on Build GRCh38
rs1028530418 Chr.13: 50910134-50910134 on Build GRCh38
rs376641184 Chr.13: 50910278-50910278 on Build GRCh38
rs908023325 Chr.13: 50910326-50910326 on Build GRCh38
rs562869389 Chr.13: 50910052-50910052 on Build GRCh38
rs757714442 Chr.13: 50910186-50910186 on Build GRCh38
rs1044215374 Chr.13: 50910305-50910305 on Build GRCh38
rs764223334 Chr.13: 50910127-50910127 on Build GRCh38
rs1022819462 Chr.13: 50909990-50909990 on Build GRCh38
rs996973016 Chr.13: 50909934-50909934 on Build GRCh38
rs770526467 Chr.13: 50910260-50910260 on Build GRCh38
rs900876937 Chr.13: 50910116-50910116 on Build GRCh38
rs529317356 Chr.13: 50910001-50910001 on Build GRCh38
rs886041377 Chr.13: 50910079-50910079 on Build GRCh38
rs990826923 Chr.13: 50910265-50910265 on Build GRCh38
rs934317454 Chr.13: 50910211-50910211 on Build GRCh38
rs886695839 Chr.13: 50910074-50910074 on Build GRCh38
rs1028060532 Chr.13: 50909955-50909955 on Build GRCh38
rs1053270229 Chr.13: 50909964-50909964 on Build GRCh38
rs957954956 Chr.13: 50910372-50910388 on Build GRCh38
rs988134864 Chr.13: 50910117-50910117 on Build GRCh38
rs9535526 Chr.13: 50910309-50910309 on Build GRCh38
rs535141690 Chr.13: 50910280-50910280 on Build GRCh38
rs1002744815 Chr.13: 50910313-50910313 on Build GRCh38
rs957777357 Chr.13: 50910148-50910148 on Build GRCh38
rs551573692 Chr.13: 50910096-50910096 on Build GRCh38
rs765552524 Chr.13: 50910068-50910068 on Build GRCh38
rs1011394240 Chr.13: 50909977-50909977 on Build GRCh38
rs549441721 Chr.13: 50910012-50910012 on Build GRCh38
rs1049363684 Chr.13: 50910389-50910389 on Build GRCh38
rs920685572 Chr.13: 50910063-50910063 on Build GRCh38
rs982736660 Chr.13: 50910108-50910108 on Build GRCh38
rs959674825 Chr.13: 50910244-50910244 on Build GRCh38
rs905756680 Chr.13: 50910004-50910004 on Build GRCh38
rs900760947 Chr.13: 50910362-50910362 on Build GRCh38
rs2274069 Chr.13: 50910220-50910220 on Build GRCh38
rs571566229 Chr.13: 50910115-50910115 on Build GRCh38
rs531904107 Chr.13: 50910083-50910083 on Build GRCh38
rs184573722 Chr.13: 50909965-50909965 on Build GRCh38
rs893563975 Chr.13: 50910396-50910396 on Build GRCh38
rs1035663072 Chr.13: 50910028-50910028 on Build GRCh38
rs575268009 Chr.13: 50910371-50910371 on Build GRCh38
rs937980132 Chr.13: 50910056-50910056 on Build GRCh38
rs553446261 Chr.13: 50909960-50909960 on Build GRCh38
rs1020774673 Chr.13: 50910200-50910200 on Build GRCh38
rs534222775 Chr.13: 50910218-50910218 on Build GRCh38
rs1004925162 Chr.13: 50910036-50910036 on Build GRCh38
rs933586659 Chr.13: 50909947-50909947 on Build GRCh38
rs950769265 Chr.13: 50910034-50910034 on Build GRCh38
rs929922905 Chr.13: 50910102-50910102 on Build GRCh38
rs996507964 Chr.13: 50910125-50910125 on Build GRCh38
rs944863640 Chr.13: 50910332-50910332 on Build GRCh38
rs920686105 Chr.13: 50910268-50910268 on Build GRCh38
rs535818770 Chr.13: 50910199-50910199 on Build GRCh38
rs966630128 Chr.13: 50910217-50910217 on Build GRCh38
rs1050759505 Chr.13: 50909953-50909953 on Build GRCh38
rs745585144 Chr.13: 50910358-50910358 on Build GRCh38
rs763103642 Chr.13: 50910122-50910122 on Build GRCh38
rs1016231511 Chr.13: 50910363-50910363 on Build GRCh38
rs982127083 Chr.13: 50910051-50910051 on Build GRCh38
rs758877268 Chr.13: 50910128-50910128 on Build GRCh38
rs79367981 Chr.13: 50910141-50910141 on Build GRCh38
rs1016285639 Chr.13: 50910049-50910049 on Build GRCh38
rs886383415 Chr.13: 50910361-50910361 on Build GRCh38
rs886050289 Chr.13: 50909970-50909970 on Build GRCh38
rs1055639616 Chr.13: 50910060-50910060 on Build GRCh38
rs1009081674 Chr.13: 50910077-50910077 on Build GRCh38

View Full Product Details