Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 50930541-50931053 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RNASEH2B
Entrez Gene ID
79621
Gene Name
ribonuclease H2 subunit B
Gene Aliases
AGS2, DLEU8
UniGene
Hs.306291
Gene Chromosome Location
Chr.13: 50909678-50970462 on Build GRCh38
Transcripts
NM_001142279.2 NM_024570.3 XM_006719867.3 XM_011535229.1 XM_011535230.2 XM_011535231.2 XM_011535233.2 XM_017020747.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4481442 c.250C>A p.L84I Chr.13: 50930688-50930688 on Build GRCh38
3728274 c.310G>A p.A104T Chr.13: 50930748-50930748 on Build GRCh38
1706782 c.318G>T p.K106N Chr.13: 50930756-50930756 on Build GRCh38
5481137 c.316A>G p.K106E Chr.13: 50930754-50930754 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs372703323 Chr.13: 50930774-50930774 on Build GRCh38
rs1010999844 Chr.13: 50930929-50930929 on Build GRCh38
rs575523187 Chr.13: 50930866-50930866 on Build GRCh38
rs893984122 Chr.13: 50930904-50930904 on Build GRCh38
rs112929301 Chr.13: 50930639-50930639 on Build GRCh38
rs770186213 Chr.13: 50930808-50930808 on Build GRCh38
rs959811072 Chr.13: 50930709-50930709 on Build GRCh38
rs755936337 Chr.13: 50930755-50930755 on Build GRCh38
rs77931005 Chr.13: 50930695-50930695 on Build GRCh38
rs908434668 Chr.13: 50930834-50930834 on Build GRCh38
rs751197802 Chr.13: 50931023-50931024 on Build GRCh38
rs1049972894 Chr.13: 50931009-50931009 on Build GRCh38
rs1028480338 Chr.13: 50930584-50930584 on Build GRCh38
rs375826744 Chr.13: 50930636-50930636 on Build GRCh38
rs1049760095 Chr.13: 50930901-50930901 on Build GRCh38
rs958197309 Chr.13: 50930593-50930593 on Build GRCh38
rs898252081 Chr.13: 50930647-50930647 on Build GRCh38
rs766207888 Chr.13: 50930767-50930767 on Build GRCh38
rs781518432 Chr.13: 50930705-50930706 on Build GRCh38
rs530546853 Chr.13: 50930571-50930571 on Build GRCh38
rs534345984 Chr.13: 50930952-50930952 on Build GRCh38
rs770203126 Chr.13: 50930707-50930707 on Build GRCh38
rs984052151 Chr.13: 50930817-50930817 on Build GRCh38
rs201078944 Chr.13: 50930752-50930752 on Build GRCh38
rs76158094 Chr.13: 50930685-50930685 on Build GRCh38
rs915751974 Chr.13: 50930766-50930766 on Build GRCh38
rs968826871 Chr.13: 50930732-50930732 on Build GRCh38
rs143523027 Chr.13: 50930688-50930688 on Build GRCh38
rs200935335 Chr.13: 50930777-50930777 on Build GRCh38
rs901159162 Chr.13: 50930846-50930846 on Build GRCh38
rs763646959 Chr.13: 50930715-50930715 on Build GRCh38
rs750291347 Chr.13: 50930649-50930649 on Build GRCh38
rs775987185 Chr.13: 50930811-50930811 on Build GRCh38
rs752778588 Chr.13: 50930662-50930662 on Build GRCh38
rs908126088 Chr.13: 50930860-50930860 on Build GRCh38
rs113695025 Chr.13: 50930648-50930648 on Build GRCh38
rs939685384 Chr.13: 50930598-50930598 on Build GRCh38
rs1057517815 Chr.13: 50930675-50930675 on Build GRCh38
rs774041308 Chr.13: 50930726-50930726 on Build GRCh38
rs1555625 Chr.13: 50930872-50930872 on Build GRCh38
rs939912071 Chr.13: 50930837-50930837 on Build GRCh38
rs747061909 Chr.13: 50930689-50930689 on Build GRCh38
rs889588886 Chr.13: 50931011-50931011 on Build GRCh38
rs763670350 Chr.13: 50930635-50930635 on Build GRCh38
rs1036691577 Chr.13: 50930883-50930883 on Build GRCh38
rs374098961 Chr.13: 50930711-50930711 on Build GRCh38
rs555661026 Chr.13: 50930853-50930853 on Build GRCh38
rs188520724 Chr.13: 50930654-50930654 on Build GRCh38
rs370521248 Chr.13: 50930807-50930807 on Build GRCh38
rs758688261 Chr.13: 50930772-50930772 on Build GRCh38
rs761380890 Chr.13: 50930742-50930742 on Build GRCh38
rs902793977 Chr.13: 50930959-50930959 on Build GRCh38
rs767086382 Chr.13: 50930746-50930746 on Build GRCh38
rs1019503838 Chr.13: 50930671-50930671 on Build GRCh38
rs768598289 Chr.13: 50930654-50930655 on Build GRCh38
rs757362657 Chr.13: 50930779-50930779 on Build GRCh38
rs1041079228 Chr.13: 50930838-50930838 on Build GRCh38
rs758538142 Chr.13: 50930678-50930678 on Build GRCh38
rs746338829 Chr.13: 50930706-50930706 on Build GRCh38
rs777149786 Chr.13: 50930701-50930701 on Build GRCh38
rs753571443 Chr.13: 50930770-50930770 on Build GRCh38
rs889639795 Chr.13: 50930650-50930650 on Build GRCh38
rs181556239 Chr.13: 50930659-50930659 on Build GRCh38
rs748404966 Chr.13: 50930674-50930674 on Build GRCh38

View Full Product Details