Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 50934725-50935217 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RNASEH2B
Entrez Gene ID
79621
Gene Name
ribonuclease H2 subunit B
Gene Aliases
AGS2, DLEU8
UniGene
Hs.306291
Gene Chromosome Location
Chr.13: 50909678-50970462 on Build GRCh38
Transcripts
NM_001142279.2 NM_024570.3 XM_006719867.3 XM_011535229.1 XM_011535230.2 XM_011535231.2 XM_011535233.2 XM_017020747.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
947929 c.374G>A p.C125Y Chr.13: 50934937-50934937 on Build GRCh38
6272987 c.384G>T p.L128F Chr.13: 50934947-50934947 on Build GRCh38
7203781 c.423G>A p.V141V Chr.13: 50934986-50934986 on Build GRCh38
947928 c.323G>A p.G108E Chr.13: 50934886-50934886 on Build GRCh38
7212312 c.322-10_322-9insT p.? Chr.13: 50934875-50934876 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759273327 Chr.13: 50934923-50934923 on Build GRCh38
rs765033294 Chr.13: 50935043-50935043 on Build GRCh38
rs963263480 Chr.13: 50935193-50935193 on Build GRCh38
rs370841549 Chr.13: 50935042-50935042 on Build GRCh38
rs769439629 Chr.13: 50935102-50935102 on Build GRCh38
rs41292768 Chr.13: 50934831-50934831 on Build GRCh38
rs1038792789 Chr.13: 50934807-50934807 on Build GRCh38
rs999474934 Chr.13: 50934893-50934893 on Build GRCh38
rs373276336 Chr.13: 50934856-50934856 on Build GRCh38
rs786205483 Chr.13: 50934919-50934919 on Build GRCh38
rs372326476 Chr.13: 50934837-50934837 on Build GRCh38
rs555874041 Chr.13: 50934841-50934841 on Build GRCh38
rs764980431 Chr.13: 50934924-50934924 on Build GRCh38
rs540876800 Chr.13: 50935146-50935146 on Build GRCh38
rs770781962 Chr.13: 50934875-50934876 on Build GRCh38
rs143699275 Chr.13: 50934812-50934812 on Build GRCh38
rs936144809 Chr.13: 50935088-50935088 on Build GRCh38
rs1014506026 Chr.13: 50935109-50935109 on Build GRCh38
rs781310346 Chr.13: 50934938-50934938 on Build GRCh38
rs780675862 Chr.13: 50934829-50934829 on Build GRCh38
rs776467802 Chr.13: 50935028-50935028 on Build GRCh38
rs61284187 Chr.13: 50935190-50935190 on Build GRCh38
rs906157484 Chr.13: 50935161-50935161 on Build GRCh38
rs752480002 Chr.13: 50935044-50935044 on Build GRCh38
rs951698581 Chr.13: 50935151-50935151 on Build GRCh38
rs80087649 Chr.13: 50935000-50935000 on Build GRCh38
rs780685003 Chr.13: 50935060-50935060 on Build GRCh38
rs754434417 Chr.13: 50934992-50934992 on Build GRCh38
rs906676856 Chr.13: 50935115-50935115 on Build GRCh38
rs751788888 Chr.13: 50934928-50934928 on Build GRCh38
rs908905660 Chr.13: 50934777-50934777 on Build GRCh38
rs756682554 Chr.13: 50934950-50934950 on Build GRCh38
rs892070504 Chr.13: 50935106-50935106 on Build GRCh38
rs200535256 Chr.13: 50934991-50934991 on Build GRCh38
rs941780807 Chr.13: 50934802-50934802 on Build GRCh38
rs772788079 Chr.13: 50934883-50934883 on Build GRCh38
rs769173136 Chr.13: 50934966-50934966 on Build GRCh38
rs943496819 Chr.13: 50934981-50934981 on Build GRCh38
rs767139201 Chr.13: 50934882-50934882 on Build GRCh38
rs772819265 Chr.13: 50935004-50935004 on Build GRCh38
rs146774907 Chr.13: 50935169-50935169 on Build GRCh38
rs770427871 Chr.13: 50935026-50935026 on Build GRCh38
rs749750512 Chr.13: 50934958-50934958 on Build GRCh38
rs759455109 Chr.13: 50935030-50935030 on Build GRCh38
rs760734846 Chr.13: 50934887-50934887 on Build GRCh38
rs748279384 Chr.13: 50934874-50934875 on Build GRCh38
rs970497502 Chr.13: 50934782-50934782 on Build GRCh38
rs1046134818 Chr.13: 50935127-50935127 on Build GRCh38
rs1001949505 Chr.13: 50934806-50934806 on Build GRCh38
rs760082711 Chr.13: 50935022-50935022 on Build GRCh38
rs780398368 Chr.13: 50934957-50934957 on Build GRCh38
rs199935006 Chr.13: 50934970-50934970 on Build GRCh38
rs571873512 Chr.13: 50935049-50935049 on Build GRCh38
rs757415855 Chr.13: 50934933-50934933 on Build GRCh38
rs766196712 Chr.13: 50934911-50934911 on Build GRCh38
rs753716315 Chr.13: 50934915-50934915 on Build GRCh38
rs569343532 Chr.13: 50934939-50934939 on Build GRCh38
rs866597039 Chr.13: 50935107-50935107 on Build GRCh38
rs558250799 Chr.13: 50935048-50935048 on Build GRCh38
rs761515995 Chr.13: 50934880-50934880 on Build GRCh38
rs568672395 Chr.13: 50934824-50934824 on Build GRCh38
rs140076332 Chr.13: 50934810-50934810 on Build GRCh38
rs893847811 Chr.13: 50935007-50935007 on Build GRCh38
rs78705382 Chr.13: 50934975-50934975 on Build GRCh38
rs747641441 Chr.13: 50935075-50935075 on Build GRCh38

View Full Product Details