Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 89777986-89778496 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MESP2
Entrez Gene ID
145873
Gene Name
mesoderm posterior bHLH transcription factor 2
Gene Aliases
SCDO2, bHLHc6
UniGene
Hs.37311
Gene Chromosome Location
Chr.15: 89776358-89778754 on Build GRCh38
Transcripts
NM_001039958.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4647864 c.931T>G p.S311A Chr.15: 89778071-89778071 on Build GRCh38
1749371 c.1181G>A p.G394D Chr.15: 89778321-89778321 on Build GRCh38
471224 c.1180G>A p.G394S Chr.15: 89778320-89778320 on Build GRCh38
6711538 c.1081G>A p.A361T Chr.15: 89778221-89778221 on Build GRCh38
4804213 c.1121G>T p.G374V Chr.15: 89778261-89778261 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763376959 Chr.15: 89778214-89778214 on Build GRCh38
rs201495246 Chr.15: 89778196-89778196 on Build GRCh38
rs372062720 Chr.15: 89778339-89778339 on Build GRCh38
rs762014488 Chr.15: 89778220-89778220 on Build GRCh38
rs751338786 Chr.15: 89778372-89778372 on Build GRCh38
rs770241465 Chr.15: 89778311-89778311 on Build GRCh38
rs886051547 Chr.15: 89778468-89778469 on Build GRCh38
rs568196227 Chr.15: 89778471-89778471 on Build GRCh38
rs546107097 Chr.15: 89778212-89778212 on Build GRCh38
rs923308382 Chr.15: 89778278-89778279 on Build GRCh38
rs777196250 Chr.15: 89778210-89778210 on Build GRCh38
rs375171689 Chr.15: 89778331-89778331 on Build GRCh38
rs760177391 Chr.15: 89778120-89778120 on Build GRCh38
rs746574351 Chr.15: 89778199-89778199 on Build GRCh38
rs752972579 Chr.15: 89778145-89778145 on Build GRCh38
rs751673727 Chr.15: 89778072-89778072 on Build GRCh38
rs768395213 Chr.15: 89778019-89778019 on Build GRCh38
rs775941947 Chr.15: 89778199-89778200 on Build GRCh38
rs771471316 Chr.15: 89778125-89778125 on Build GRCh38
rs182411386 Chr.15: 89778432-89778432 on Build GRCh38
rs768139078 Chr.15: 89778311-89778312 on Build GRCh38
rs772586292 Chr.15: 89778153-89778153 on Build GRCh38
rs914363098 Chr.15: 89778404-89778404 on Build GRCh38
rs547275737 Chr.15: 89778376-89778376 on Build GRCh38
rs768285654 Chr.15: 89778106-89778106 on Build GRCh38
rs1056747683 Chr.15: 89778452-89778452 on Build GRCh38
rs897440052 Chr.15: 89778160-89778160 on Build GRCh38
rs905495693 Chr.15: 89778104-89778104 on Build GRCh38
rs761113695 Chr.15: 89778039-89778039 on Build GRCh38
rs925442555 Chr.15: 89778069-89778069 on Build GRCh38
rs758225691 Chr.15: 89778352-89778352 on Build GRCh38
rs190964237 Chr.15: 89778417-89778417 on Build GRCh38
rs770301033 Chr.15: 89778203-89778203 on Build GRCh38
rs530131979 Chr.15: 89778359-89778359 on Build GRCh38
rs752818594 Chr.15: 89778051-89778051 on Build GRCh38
rs780062393 Chr.15: 89778158-89778158 on Build GRCh38
rs745376223 Chr.15: 89778309-89778309 on Build GRCh38
rs774720315 Chr.15: 89778318-89778318 on Build GRCh38
rs754298708 Chr.15: 89778043-89778043 on Build GRCh38
rs761831896 Chr.15: 89778435-89778435 on Build GRCh38
rs749949151 Chr.15: 89778383-89778383 on Build GRCh38
rs752531314 Chr.15: 89778266-89778266 on Build GRCh38
rs754930000 Chr.15: 89778244-89778244 on Build GRCh38
rs529473935 Chr.15: 89778363-89778363 on Build GRCh38
rs749691994 Chr.15: 89778315-89778315 on Build GRCh38
rs746629107 Chr.15: 89778298-89778298 on Build GRCh38
rs777428484 Chr.15: 89778280-89778280 on Build GRCh38
rs746380436 Chr.15: 89778145-89778146 on Build GRCh38
rs776583904 Chr.15: 89778424-89778424 on Build GRCh38
rs544384142 Chr.15: 89778334-89778334 on Build GRCh38
rs1044843208 Chr.15: 89778118-89778118 on Build GRCh38
rs765211311 Chr.15: 89778342-89778342 on Build GRCh38
rs767430602 Chr.15: 89778221-89778221 on Build GRCh38
rs556469683 Chr.15: 89778457-89778457 on Build GRCh38
rs776116878 Chr.15: 89778314-89778314 on Build GRCh38
rs200678001 Chr.15: 89778293-89778293 on Build GRCh38
rs896844056 Chr.15: 89778456-89778456 on Build GRCh38
rs879095532 Chr.15: 89778154-89778154 on Build GRCh38
rs564292487 Chr.15: 89778213-89778213 on Build GRCh38
rs917606111 Chr.15: 89778082-89778082 on Build GRCh38
rs765547733 Chr.15: 89778048-89778048 on Build GRCh38
rs3840032 Chr.15: 89778464-89778465 on Build GRCh38
rs556175245 Chr.15: 89778044-89778044 on Build GRCh38
rs749029705 Chr.15: 89778016-89778016 on Build GRCh38
rs543667424 Chr.15: 89778294-89778296 on Build GRCh38
rs199514997 Chr.15: 89778184-89778184 on Build GRCh38
rs779823676 Chr.15: 89778081-89778081 on Build GRCh38
rs376942190 Chr.15: 89778142-89778142 on Build GRCh38
rs570676731 Chr.15: 89778021-89778021 on Build GRCh38
rs765449941 Chr.15: 89778144-89778144 on Build GRCh38
rs370227491 Chr.15: 89778157-89778157 on Build GRCh38
rs755600035 Chr.15: 89778384-89778384 on Build GRCh38
rs761885039 Chr.15: 89778330-89778330 on Build GRCh38
rs780864633 Chr.15: 89778380-89778380 on Build GRCh38
rs758552055 Chr.15: 89778052-89778052 on Build GRCh38
rs754807279 Chr.15: 89778176-89778176 on Build GRCh38
rs187988937 Chr.15: 89778441-89778441 on Build GRCh38
rs758115773 Chr.15: 89778190-89778190 on Build GRCh38
rs187680280 Chr.15: 89778343-89778343 on Build GRCh38
rs777939697 Chr.15: 89778065-89778065 on Build GRCh38
rs374475842 Chr.15: 89778167-89778167 on Build GRCh38
rs771586079 Chr.15: 89778115-89778115 on Build GRCh38
rs756801528 Chr.15: 89778303-89778303 on Build GRCh38
rs766860893 Chr.15: 89778040-89778040 on Build GRCh38
rs71647810 Chr.15: 89778192-89778192 on Build GRCh38
rs776986584 Chr.15: 89778127-89778127 on Build GRCh38
rs774665542 Chr.15: 89778218-89778218 on Build GRCh38
rs765218369 Chr.15: 89778262-89778262 on Build GRCh38
rs976784144 Chr.15: 89778348-89778348 on Build GRCh38
rs747926651 Chr.15: 89778187-89778187 on Build GRCh38
rs753827969 Chr.15: 89778229-89778229 on Build GRCh38
rs778659855 Chr.15: 89778107-89778107 on Build GRCh38
rs763321859 Chr.15: 89778150-89778150 on Build GRCh38
rs756185244 Chr.15: 89778080-89778080 on Build GRCh38
rs558760540 Chr.15: 89778020-89778020 on Build GRCh38
rs757634059 Chr.15: 89778088-89778089 on Build GRCh38
rs372981270 Chr.15: 89778116-89778116 on Build GRCh38
rs376465704 Chr.15: 89778217-89778217 on Build GRCh38
rs780621098 Chr.15: 89778306-89778306 on Build GRCh38
rs771530948 Chr.15: 89778025-89778025 on Build GRCh38
rs779274067 Chr.15: 89778142-89778143 on Build GRCh38
rs527993697 Chr.15: 89778407-89778407 on Build GRCh38
rs1018363425 Chr.15: 89778328-89778328 on Build GRCh38
rs537567063 Chr.15: 89778031-89778031 on Build GRCh38
rs764403618 Chr.15: 89778151-89778151 on Build GRCh38
rs752665246 Chr.15: 89778097-89778097 on Build GRCh38
rs758170634 Chr.15: 89778267-89778267 on Build GRCh38
rs747732638 Chr.15: 89778113-89778113 on Build GRCh38

View Full Product Details