Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 1351508-1352010 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GNPTG
Entrez Gene ID
84572
Gene Name
N-acetylglucosamine-1-phosphate transferase gamma subunit
Gene Aliases
C16orf27, GNPTAG, LP2537, RJD9
UniGene
Hs.241575
Gene Chromosome Location
Chr.16: 1351899-1363351 on Build GRCh38
Transcripts
NM_032520.4 XM_017023782.1 NM_001001410.2

Gene Symbol
TSR3
Entrez Gene ID
115939
Gene Name
TSR3, acp transferase ribosome maturation factor
Gene Aliases
C16orf42
UniGene
Hs.134846
Gene Chromosome Location
Chr.16: 1349240-1351872 on Build GRCh38
Transcripts
NM_032520.4 XM_017023782.1 NM_001001410.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6349664 c.52C>T p.R18W Chr.16: 1351753-1351753 on Build GRCh38
5657723 c.25G>T p.G9W Chr.16: 1351780-1351780 on Build GRCh38
6364781 c.173G>A p.C58Y Chr.16: 1351538-1351538 on Build GRCh38
6555328 c.165G>A p.A55A Chr.16: 1351546-1351546 on Build GRCh38
6421685 c.118G>A p.V40M Chr.16: 1351593-1351593 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1054163328 Chr.16: 1351759-1351759 on Build GRCh38
rs899575751 Chr.16: 1351546-1351546 on Build GRCh38
rs888194280 Chr.16: 1351777-1351777 on Build GRCh38
rs536513000 Chr.16: 1351731-1351731 on Build GRCh38
rs905778790 Chr.16: 1351918-1351933 on Build GRCh38
rs1004055677 Chr.16: 1351583-1351583 on Build GRCh38
rs760140489 Chr.16: 1351765-1351765 on Build GRCh38
rs986694444 Chr.16: 1351955-1351955 on Build GRCh38
rs763764801 Chr.16: 1351974-1351974 on Build GRCh38
rs8062301 Chr.16: 1351795-1351795 on Build GRCh38
rs547587708 Chr.16: 1351701-1351701 on Build GRCh38
rs1016193943 Chr.16: 1351924-1351924 on Build GRCh38
rs756910244 Chr.16: 1351977-1351977 on Build GRCh38
rs1034850148 Chr.16: 1351826-1351826 on Build GRCh38
rs111834368 Chr.16: 1351766-1351766 on Build GRCh38
rs1006329439 Chr.16: 1351922-1351922 on Build GRCh38
rs942360505 Chr.16: 1351744-1351744 on Build GRCh38
rs1041664887 Chr.16: 1351906-1351906 on Build GRCh38
rs567583327 Chr.16: 1351703-1351703 on Build GRCh38
rs532236241 Chr.16: 1351984-1351984 on Build GRCh38
rs972306295 Chr.16: 1351841-1351841 on Build GRCh38
rs1003116954 Chr.16: 1351908-1351908 on Build GRCh38
rs972447741 Chr.16: 1351975-1351975 on Build GRCh38
rs756390327 Chr.16: 1351530-1351530 on Build GRCh38
rs565020861 Chr.16: 1351811-1351811 on Build GRCh38
rs932328410 Chr.16: 1351761-1351761 on Build GRCh38
rs1055666228 Chr.16: 1351911-1351911 on Build GRCh38
rs914153555 Chr.16: 1351732-1351732 on Build GRCh38
rs1040726547 Chr.16: 1351545-1351545 on Build GRCh38
rs981543236 Chr.16: 1351590-1351590 on Build GRCh38
rs886051657 Chr.16: 1351924-1351924 on Build GRCh38
rs1013581887 Chr.16: 1351618-1351618 on Build GRCh38
rs980855523 Chr.16: 1351679-1351679 on Build GRCh38
rs923463438 Chr.16: 1351714-1351714 on Build GRCh38
rs909452032 Chr.16: 1351649-1351649 on Build GRCh38
rs893881176 Chr.16: 1351927-1351927 on Build GRCh38
rs988271287 Chr.16: 1351724-1351724 on Build GRCh38
rs772239803 Chr.16: 1351991-1351991 on Build GRCh38
rs942495145 Chr.16: 1351874-1351874 on Build GRCh38
rs28541713 Chr.16: 1351844-1351844 on Build GRCh38
rs772436988 Chr.16: 1351646-1351646 on Build GRCh38
rs964111307 Chr.16: 1351960-1351960 on Build GRCh38
rs886051658 Chr.16: 1351971-1351971 on Build GRCh38
rs1010066590 Chr.16: 1351560-1351560 on Build GRCh38
rs867217705 Chr.16: 1351973-1351973 on Build GRCh38
rs543660760 Chr.16: 1351978-1351978 on Build GRCh38
rs955019316 Chr.16: 1351949-1351949 on Build GRCh38
rs1021014743 Chr.16: 1351943-1351943 on Build GRCh38
rs1054511796 Chr.16: 1351885-1351885 on Build GRCh38
rs907048726 Chr.16: 1351789-1351789 on Build GRCh38
rs928528974 Chr.16: 1351988-1351988 on Build GRCh38
rs776466792 Chr.16: 1351743-1351743 on Build GRCh38
rs562102467 Chr.16: 1351571-1351571 on Build GRCh38
rs910957824 Chr.16: 1351869-1351869 on Build GRCh38
rs1041550517 Chr.16: 1351784-1351784 on Build GRCh38
rs959983428 Chr.16: 1351613-1351613 on Build GRCh38
rs945918643 Chr.16: 1351897-1351897 on Build GRCh38
rs1044660992 Chr.16: 1351992-1351992 on Build GRCh38
rs556601696 Chr.16: 1351742-1351742 on Build GRCh38
rs906996362 Chr.16: 1351907-1351907 on Build GRCh38
rs944329193 Chr.16: 1351733-1351733 on Build GRCh38
rs551184048 Chr.16: 1351640-1351640 on Build GRCh38
rs888263919 Chr.16: 1351572-1351572 on Build GRCh38
rs779505508 Chr.16: 1351985-1351985 on Build GRCh38
rs541284873 Chr.16: 1351926-1351926 on Build GRCh38
rs765220411 Chr.16: 1351938-1351938 on Build GRCh38
rs893047539 Chr.16: 1351760-1351760 on Build GRCh38
rs1034326211 Chr.16: 1351626-1351626 on Build GRCh38
rs996639866 Chr.16: 1351551-1351551 on Build GRCh38
rs971453741 Chr.16: 1351589-1351589 on Build GRCh38
rs969858614 Chr.16: 1351636-1351636 on Build GRCh38
rs868174441 Chr.16: 1351825-1351825 on Build GRCh38
rs59225734 Chr.16: 1351929-1351944 on Build GRCh38
rs780629813 Chr.16: 1351550-1351550 on Build GRCh38
rs754225455 Chr.16: 1351982-1351982 on Build GRCh38
rs1036898400 Chr.16: 1351803-1351803 on Build GRCh38
rs539165958 Chr.16: 1351833-1351833 on Build GRCh38
rs1014203471 Chr.16: 1351823-1351823 on Build GRCh38
rs752212692 Chr.16: 1351944-1351944 on Build GRCh38
rs749807152 Chr.16: 1351556-1351556 on Build GRCh38
rs772713427 Chr.16: 1351658-1351658 on Build GRCh38
rs893099979 Chr.16: 1351549-1351549 on Build GRCh38
rs549749503 Chr.16: 1351684-1351684 on Build GRCh38
rs865811325 Chr.16: 1351702-1351702 on Build GRCh38
rs958857563 Chr.16: 1351827-1351827 on Build GRCh38
rs527473522 Chr.16: 1351686-1351686 on Build GRCh38
rs570065824 Chr.16: 1351832-1351832 on Build GRCh38
rs958198845 Chr.16: 1351712-1351712 on Build GRCh38
rs373656284 Chr.16: 1351627-1351627 on Build GRCh38
rs962248757 Chr.16: 1351837-1351837 on Build GRCh38
rs954881938 Chr.16: 1351854-1351854 on Build GRCh38
rs993673873 Chr.16: 1351932-1351932 on Build GRCh38
rs8063740 Chr.16: 1351562-1351562 on Build GRCh38
rs894537198 Chr.16: 1351916-1351916 on Build GRCh38
rs28654239 Chr.16: 1351894-1351894 on Build GRCh38
rs1048108633 Chr.16: 1351769-1351769 on Build GRCh38
rs574801192 Chr.16: 1351976-1351976 on Build GRCh38
rs914207699 Chr.16: 1351972-1351972 on Build GRCh38
rs1032389243 Chr.16: 1351829-1351829 on Build GRCh38
rs986356876 Chr.16: 1351856-1351856 on Build GRCh38
rs895722963 Chr.16: 1351806-1351806 on Build GRCh38
rs554707396 Chr.16: 1351962-1351962 on Build GRCh38
rs886051656 Chr.16: 1351906-1351906 on Build GRCh38
rs1017797000 Chr.16: 1351956-1351956 on Build GRCh38
rs748226660 Chr.16: 1351643-1351643 on Build GRCh38
rs989913303 Chr.16: 1351971-1351971 on Build GRCh38
rs977124651 Chr.16: 1351986-1351986 on Build GRCh38
rs1051298505 Chr.16: 1351726-1351726 on Build GRCh38
rs1004044382 Chr.16: 1351787-1351787 on Build GRCh38
rs748243075 Chr.16: 1351990-1351990 on Build GRCh38
rs933717447 Chr.16: 1351725-1351725 on Build GRCh38
rs867116818 Chr.16: 1351836-1351836 on Build GRCh38
rs1015478462 Chr.16: 1351598-1351598 on Build GRCh38

View Full Product Details