Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 1362372-1362866 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GNPTG
Entrez Gene ID
84572
Gene Name
N-acetylglucosamine-1-phosphate transferase gamma subunit
Gene Aliases
C16orf27, GNPTAG, LP2537, RJD9
UniGene
Hs.241575
Gene Chromosome Location
Chr.16: 1351899-1363351 on Build GRCh38
Transcripts
NM_032520.4 XM_017023782.1 XM_017023783.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6672464 c.611G>A p.G204D Chr.16: 1362612-1362612 on Build GRCh38
4058425 c.751G>T p.E251* Chr.16: 1362834-1362834 on Build GRCh38
2143663 c.699T>C p.G233G Chr.16: 1362700-1362700 on Build GRCh38
701931 c.606C>A p.P202P Chr.16: 1362531-1362531 on Build GRCh38
2148936 c.603C>T p.T201T Chr.16: 1362528-1362528 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748861038 Chr.16: 1362680-1362680 on Build GRCh38
rs367672512 Chr.16: 1362514-1362514 on Build GRCh38
rs749314645 Chr.16: 1362499-1362499 on Build GRCh38
rs11554986 Chr.16: 1362458-1362458 on Build GRCh38
rs746879107 Chr.16: 1362487-1362488 on Build GRCh38
rs753661059 Chr.16: 1362766-1362766 on Build GRCh38
rs150965148 Chr.16: 1362685-1362685 on Build GRCh38
rs761527949 Chr.16: 1362644-1362644 on Build GRCh38
rs966758681 Chr.16: 1362682-1362682 on Build GRCh38
rs766050367 Chr.16: 1362405-1362405 on Build GRCh38
rs371060844 Chr.16: 1362748-1362748 on Build GRCh38
rs775527344 Chr.16: 1362679-1362679 on Build GRCh38
rs368698403 Chr.16: 1362556-1362556 on Build GRCh38
rs532275192 Chr.16: 1362703-1362703 on Build GRCh38
rs750752675 Chr.16: 1362530-1362530 on Build GRCh38
rs117489832 Chr.16: 1362548-1362548 on Build GRCh38
rs759248626 Chr.16: 1362473-1362473 on Build GRCh38
rs201844106 Chr.16: 1362652-1362652 on Build GRCh38
rs749582831 Chr.16: 1362448-1362448 on Build GRCh38
rs990613310 Chr.16: 1362702-1362702 on Build GRCh38
rs193302858 Chr.16: 1362640-1362640 on Build GRCh38
rs180901866 Chr.16: 1362567-1362567 on Build GRCh38
rs142548931 Chr.16: 1362666-1362666 on Build GRCh38
rs112266724 Chr.16: 1362522-1362522 on Build GRCh38
rs778168070 Chr.16: 1362420-1362420 on Build GRCh38
rs969101852 Chr.16: 1362759-1362760 on Build GRCh38
rs781057196 Chr.16: 1362558-1362558 on Build GRCh38
rs745410307 Chr.16: 1362560-1362560 on Build GRCh38
rs200239398 Chr.16: 1362770-1362770 on Build GRCh38
rs544780390 Chr.16: 1362544-1362544 on Build GRCh38
rs768997138 Chr.16: 1362450-1362450 on Build GRCh38
rs545803225 Chr.16: 1362404-1362404 on Build GRCh38
rs780289639 Chr.16: 1362806-1362806 on Build GRCh38
rs575715803 Chr.16: 1362534-1362535 on Build GRCh38
rs193302855 Chr.16: 1362609-1362609 on Build GRCh38
rs750278694 Chr.16: 1362578-1362584 on Build GRCh38
rs770850105 Chr.16: 1362521-1362521 on Build GRCh38
rs761837002 Chr.16: 1362754-1362754 on Build GRCh38
rs769398814 Chr.16: 1362562-1362562 on Build GRCh38
rs773331398 Chr.16: 1362825-1362825 on Build GRCh38
rs764405882 Chr.16: 1362661-1362661 on Build GRCh38
rs143490078 Chr.16: 1362508-1362508 on Build GRCh38
rs749994383 Chr.16: 1362828-1362829 on Build GRCh38
rs193302852 Chr.16: 1362533-1362534 on Build GRCh38
rs771116867 Chr.16: 1362829-1362829 on Build GRCh38
rs779149342 Chr.16: 1362510-1362510 on Build GRCh38
rs780930809 Chr.16: 1362628-1362628 on Build GRCh38
rs749567145 Chr.16: 1362560-1362569 on Build GRCh38
rs776107971 Chr.16: 1362712-1362712 on Build GRCh38
rs764889321 Chr.16: 1362526-1362526 on Build GRCh38
rs762029401 Chr.16: 1362706-1362706 on Build GRCh38
rs927888758 Chr.16: 1362516-1362516 on Build GRCh38
rs760871015 Chr.16: 1362828-1362828 on Build GRCh38
rs766055577 Chr.16: 1362831-1362834 on Build GRCh38
rs774852659 Chr.16: 1362563-1362563 on Build GRCh38
rs376709253 Chr.16: 1362591-1362591 on Build GRCh38
rs200094858 Chr.16: 1362691-1362691 on Build GRCh38
rs751946577 Chr.16: 1362426-1362426 on Build GRCh38
rs751373599 Chr.16: 1362613-1362613 on Build GRCh38
rs755265539 Chr.16: 1362502-1362502 on Build GRCh38
rs370884840 Chr.16: 1362431-1362431 on Build GRCh38
rs890055206 Chr.16: 1362631-1362631 on Build GRCh38
rs750155751 Chr.16: 1362668-1362668 on Build GRCh38
rs774108877 Chr.16: 1362643-1362643 on Build GRCh38
rs771844889 Chr.16: 1362692-1362692 on Build GRCh38
rs774413019 Chr.16: 1362704-1362704 on Build GRCh38
rs756225251 Chr.16: 1362526-1362527 on Build GRCh38
rs768131774 Chr.16: 1362639-1362639 on Build GRCh38
rs766167977 Chr.16: 1362715-1362715 on Build GRCh38
rs756918285 Chr.16: 1362800-1362800 on Build GRCh38
rs771470862 Chr.16: 1362645-1362645 on Build GRCh38
rs193302857 Chr.16: 1362620-1362621 on Build GRCh38
rs761072266 Chr.16: 1362575-1362575 on Build GRCh38
rs750990691 Chr.16: 1362489-1362489 on Build GRCh38
rs753529251 Chr.16: 1362408-1362408 on Build GRCh38
rs756679779 Chr.16: 1362493-1362493 on Build GRCh38
rs777140384 Chr.16: 1362694-1362694 on Build GRCh38
rs911440995 Chr.16: 1362758-1362758 on Build GRCh38
rs112925416 Chr.16: 1362632-1362632 on Build GRCh38
rs779356410 Chr.16: 1362818-1362818 on Build GRCh38
rs375490380 Chr.16: 1362549-1362549 on Build GRCh38
rs137853827 Chr.16: 1362689-1362689 on Build GRCh38
rs751216882 Chr.16: 1362798-1362798 on Build GRCh38
rs199724765 Chr.16: 1362822-1362822 on Build GRCh38
rs773159644 Chr.16: 1362460-1362460 on Build GRCh38
rs763757057 Chr.16: 1362774-1362775 on Build GRCh38
rs374564369 Chr.16: 1362698-1362698 on Build GRCh38
rs760155537 Chr.16: 1362650-1362650 on Build GRCh38
rs762827756 Chr.16: 1362713-1362713 on Build GRCh38
rs1001955796 Chr.16: 1362764-1362764 on Build GRCh38
rs745952967 Chr.16: 1362436-1362436 on Build GRCh38
rs780062523 Chr.16: 1362418-1362419 on Build GRCh38
rs376896298 Chr.16: 1362838-1362838 on Build GRCh38
rs747918283 Chr.16: 1362456-1362456 on Build GRCh38
rs193302854 Chr.16: 1362610-1362610 on Build GRCh38
rs202129898 Chr.16: 1362739-1362739 on Build GRCh38
rs776277092 Chr.16: 1362783-1362784 on Build GRCh38
rs754601335 Chr.16: 1362409-1362409 on Build GRCh38
rs767745367 Chr.16: 1362483-1362483 on Build GRCh38
rs368747296 Chr.16: 1362730-1362730 on Build GRCh38
rs141035347 Chr.16: 1362742-1362742 on Build GRCh38
rs756093689 Chr.16: 1362671-1362671 on Build GRCh38
rs893287710 Chr.16: 1362786-1362786 on Build GRCh38
rs541942383 Chr.16: 1362513-1362513 on Build GRCh38
rs146171435 Chr.16: 1362528-1362528 on Build GRCh38
rs751880564 Chr.16: 1362481-1362481 on Build GRCh38
rs757938105 Chr.16: 1362793-1362793 on Build GRCh38
rs202080062 Chr.16: 1362714-1362714 on Build GRCh38
rs747006851 Chr.16: 1362519-1362519 on Build GRCh38
rs772287534 Chr.16: 1362684-1362684 on Build GRCh38
rs982430803 Chr.16: 1362509-1362509 on Build GRCh38
rs775816256 Chr.16: 1362701-1362701 on Build GRCh38
rs969196696 Chr.16: 1362621-1362621 on Build GRCh38
rs777120261 Chr.16: 1362648-1362648 on Build GRCh38
rs778631467 Chr.16: 1362738-1362738 on Build GRCh38
rs555595061 Chr.16: 1362810-1362810 on Build GRCh38
rs138332265 Chr.16: 1362785-1362785 on Build GRCh38
rs760497948 Chr.16: 1362462-1362462 on Build GRCh38
rs761341825 Chr.16: 1362797-1362798 on Build GRCh38
rs761299690 Chr.16: 1362841-1362841 on Build GRCh38
rs746735806 Chr.16: 1362550-1362550 on Build GRCh38
rs201263537 Chr.16: 1362747-1362747 on Build GRCh38
rs774431331 Chr.16: 1362753-1362753 on Build GRCh38
rs768377103 Chr.16: 1362642-1362642 on Build GRCh38
rs372353423 Chr.16: 1362545-1362545 on Build GRCh38
rs751817263 Chr.16: 1362664-1362664 on Build GRCh38
rs193302853 Chr.16: 1362562-1362595 on Build GRCh38
rs925124634 Chr.16: 1362633-1362633 on Build GRCh38
rs759356138 Chr.16: 1362840-1362840 on Build GRCh38
rs199607037 Chr.16: 1362752-1362752 on Build GRCh38
rs531032942 Chr.16: 1362540-1362540 on Build GRCh38
rs774367224 Chr.16: 1362597-1362598 on Build GRCh38
rs773290124 Chr.16: 1362476-1362476 on Build GRCh38
rs777805362 Chr.16: 1362551-1362560 on Build GRCh38
rs748692260 Chr.16: 1362641-1362641 on Build GRCh38
rs1054675795 Chr.16: 1362783-1362783 on Build GRCh38
rs201419201 Chr.16: 1362541-1362541 on Build GRCh38
rs754489307 Chr.16: 1362684-1362684 on Build GRCh38
rs760096636 Chr.16: 1362590-1362590 on Build GRCh38
rs780352492 Chr.16: 1362495-1362495 on Build GRCh38
rs781679611 Chr.16: 1362430-1362430 on Build GRCh38
rs762888658 Chr.16: 1362658-1362658 on Build GRCh38
rs761243739 Chr.16: 1362752-1362753 on Build GRCh38
rs368890072 Chr.16: 1362755-1362755 on Build GRCh38
rs553046300 Chr.16: 1362457-1362457 on Build GRCh38
rs747594923 Chr.16: 1362741-1362741 on Build GRCh38
rs747927356 Chr.16: 1362512-1362512 on Build GRCh38
rs758242017 Chr.16: 1362830-1362831 on Build GRCh38
rs773725307 Chr.16: 1362569-1362569 on Build GRCh38
rs764994151 Chr.16: 1362810-1362811 on Build GRCh38
rs139997459 Chr.16: 1362482-1362482 on Build GRCh38
rs757003667 Chr.16: 1362624-1362624 on Build GRCh38
rs751562009 Chr.16: 1362841-1362842 on Build GRCh38
rs746271025 Chr.16: 1362636-1362637 on Build GRCh38
rs777795921 Chr.16: 1362611-1362611 on Build GRCh38
rs576693410 Chr.16: 1362399-1362399 on Build GRCh38
rs201392900 Chr.16: 1362750-1362750 on Build GRCh38
rs374454359 Chr.16: 1362444-1362444 on Build GRCh38
rs746160240 Chr.16: 1362697-1362697 on Build GRCh38
rs749914695 Chr.16: 1362402-1362402 on Build GRCh38
rs779807567 Chr.16: 1362672-1362672 on Build GRCh38
rs573130947 Chr.16: 1362771-1362771 on Build GRCh38
rs756694104 Chr.16: 1362534-1362534 on Build GRCh38
rs373976323 Chr.16: 1362824-1362824 on Build GRCh38
rs766147394 Chr.16: 1362756-1362756 on Build GRCh38
rs754928775 Chr.16: 1362731-1362731 on Build GRCh38
rs552248197 Chr.16: 1362638-1362638 on Build GRCh38
rs752714382 Chr.16: 1362605-1362605 on Build GRCh38
rs200187984 Chr.16: 1362427-1362427 on Build GRCh38
rs776105074 Chr.16: 1362463-1362463 on Build GRCh38
rs772485364 Chr.16: 1362452-1362452 on Build GRCh38
rs770037398 Chr.16: 1362439-1362439 on Build GRCh38
rs749610702 Chr.16: 1362809-1362809 on Build GRCh38
rs767026722 Chr.16: 1362578-1362578 on Build GRCh38
rs193302856 Chr.16: 1362612-1362612 on Build GRCh38
rs754437106 Chr.16: 1362581-1362581 on Build GRCh38

View Full Product Details