Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 2599047-2599538 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PDPK1
Entrez Gene ID
5170
Gene Name
3-phosphoinositide dependent protein kinase 1
Gene Aliases
PDK1, PDPK2, PDPK2P, PRO0461
UniGene
Hs.459691
Gene Chromosome Location
Chr.16: 2537964-2603190 on Build GRCh38
Transcripts
NM_001261816.1 NM_002613.4 NM_031268.5 XM_011522521.2 XM_011522523.2 XM_017023271.1 XM_017023272.1 XM_017023273.1 XM_017023274.1 XM_017023275.1 XM_017023276.1 XM_017023277.1

SNP Detail

SNP ID Chromosome Location
rs908548525 Chr.16: 2599282-2599282 on Build GRCh38
rs937283945 Chr.16: 2599111-2599111 on Build GRCh38
rs576938086 Chr.16: 2599085-2599085 on Build GRCh38
rs149315179 Chr.16: 2599245-2599247 on Build GRCh38
rs1038368343 Chr.16: 2599387-2599387 on Build GRCh38
rs971129084 Chr.16: 2599079-2599079 on Build GRCh38
rs549760673 Chr.16: 2599456-2599456 on Build GRCh38
rs1036956345 Chr.16: 2599134-2599134 on Build GRCh38
rs532266776 Chr.16: 2599501-2599501 on Build GRCh38
rs907353631 Chr.16: 2599435-2599435 on Build GRCh38
rs529898940 Chr.16: 2599432-2599432 on Build GRCh38
rs1009681908 Chr.16: 2599520-2599520 on Build GRCh38
rs1007441669 Chr.16: 2599181-2599181 on Build GRCh38
rs933912819 Chr.16: 2599233-2599233 on Build GRCh38
rs984736269 Chr.16: 2599116-2599116 on Build GRCh38
rs941366520 Chr.16: 2599126-2599126 on Build GRCh38
rs559396383 Chr.16: 2599145-2599145 on Build GRCh38
rs981312197 Chr.16: 2599084-2599084 on Build GRCh38
rs28679688 Chr.16: 2599153-2599153 on Build GRCh38
rs373772013 Chr.16: 2599473-2599473 on Build GRCh38
rs867952914 Chr.16: 2599467-2599467 on Build GRCh38
rs956314334 Chr.16: 2599509-2599509 on Build GRCh38
rs1044562746 Chr.16: 2599432-2599432 on Build GRCh38
rs967816457 Chr.16: 2599147-2599147 on Build GRCh38
rs893353568 Chr.16: 2599211-2599211 on Build GRCh38
rs1003090338 Chr.16: 2599438-2599438 on Build GRCh38
rs898608040 Chr.16: 2599389-2599389 on Build GRCh38
rs889949943 Chr.16: 2599179-2599179 on Build GRCh38
rs1034663981 Chr.16: 2599087-2599087 on Build GRCh38
rs1051727910 Chr.16: 2599176-2599176 on Build GRCh38
rs367998725 Chr.16: 2599420-2599420 on Build GRCh38
rs545861137 Chr.16: 2599120-2599120 on Build GRCh38
rs1009914699 Chr.16: 2599223-2599223 on Build GRCh38
rs867201948 Chr.16: 2599242-2599244 on Build GRCh38
rs4786295 Chr.16: 2599408-2599408 on Build GRCh38
rs1054733571 Chr.16: 2599190-2599190 on Build GRCh38
rs372067195 Chr.16: 2599476-2599476 on Build GRCh38
rs540817771 Chr.16: 2599273-2599273 on Build GRCh38
rs939974227 Chr.16: 2599353-2599353 on Build GRCh38
rs561282155 Chr.16: 2599286-2599286 on Build GRCh38
rs770202665 Chr.16: 2599090-2599090 on Build GRCh38
rs1012939731 Chr.16: 2599421-2599421 on Build GRCh38

View Full Product Details