Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 3581657-3582162 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLX4
Entrez Gene ID
84464
Gene Name
SLX4 structure-specific endonuclease subunit
Gene Aliases
BTBD12, FANCP, MUS312
UniGene
Hs.143681
Gene Chromosome Location
Chr.16: 3581181-3611598 on Build GRCh38
Transcripts
NM_032444.2 XM_011522715.2 XM_017023775.1

SNP Detail

SNP ID Chromosome Location
rs1005954025 Chr.16: 3581887-3581888 on Build GRCh38
rs896565707 Chr.16: 3581915-3581915 on Build GRCh38
rs1052504775 Chr.16: 3581894-3581894 on Build GRCh38
rs896519004 Chr.16: 3581763-3581763 on Build GRCh38
rs73505410 Chr.16: 3582090-3582090 on Build GRCh38
rs573269161 Chr.16: 3581906-3581906 on Build GRCh38
rs1026893457 Chr.16: 3581976-3581976 on Build GRCh38
rs1025661929 Chr.16: 3581975-3581977 on Build GRCh38
rs922183953 Chr.16: 3581818-3581818 on Build GRCh38
rs546917026 Chr.16: 3581690-3581690 on Build GRCh38
rs965120920 Chr.16: 3582062-3582062 on Build GRCh38
rs1052452469 Chr.16: 3581759-3581759 on Build GRCh38
rs542209079 Chr.16: 3581911-3581911 on Build GRCh38
rs923199206 Chr.16: 3582054-3582058 on Build GRCh38
rs975495836 Chr.16: 3582089-3582089 on Build GRCh38
rs911261473 Chr.16: 3581826-3581826 on Build GRCh38
rs535480497 Chr.16: 3581769-3581769 on Build GRCh38
rs78541056 Chr.16: 3581973-3581973 on Build GRCh38
rs183576223 Chr.16: 3581974-3581974 on Build GRCh38
rs975065382 Chr.16: 3581817-3581817 on Build GRCh38
rs116003727 Chr.16: 3581687-3581687 on Build GRCh38
rs942673733 Chr.16: 3581847-3581847 on Build GRCh38
rs781322530 Chr.16: 3581764-3581764 on Build GRCh38
rs1045045712 Chr.16: 3581965-3581965 on Build GRCh38
rs554146344 Chr.16: 3581871-3581871 on Build GRCh38
rs773996272 Chr.16: 3581713-3581714 on Build GRCh38
rs369843531 Chr.16: 3581968-3581968 on Build GRCh38
rs564404327 Chr.16: 3581685-3581685 on Build GRCh38
rs933276266 Chr.16: 3582055-3582058 on Build GRCh38
rs901711994 Chr.16: 3581688-3581688 on Build GRCh38
rs541410329 Chr.16: 3581684-3581684 on Build GRCh38
rs550608561 Chr.16: 3581881-3581881 on Build GRCh38
rs758604467 Chr.16: 3581904-3581904 on Build GRCh38
rs374613336 Chr.16: 3581719-3581719 on Build GRCh38
rs555535779 Chr.16: 3581975-3581975 on Build GRCh38
rs115773986 Chr.16: 3581808-3581808 on Build GRCh38
rs886252086 Chr.16: 3581878-3581878 on Build GRCh38
rs935394137 Chr.16: 3581756-3581756 on Build GRCh38
rs911269430 Chr.16: 3582041-3582052 on Build GRCh38
rs984439099 Chr.16: 3581739-3581739 on Build GRCh38
rs534067437 Chr.16: 3581859-3581859 on Build GRCh38
rs566716185 Chr.16: 3582041-3582044 on Build GRCh38
rs1015905058 Chr.16: 3581741-3581741 on Build GRCh38
rs557956614 Chr.16: 3581821-3581821 on Build GRCh38
rs949422992 Chr.16: 3581923-3581923 on Build GRCh38
rs146163310 Chr.16: 3581787-3581787 on Build GRCh38
rs989601408 Chr.16: 3582041-3582045 on Build GRCh38
rs886975991 Chr.16: 3581991-3581991 on Build GRCh38
rs530681898 Chr.16: 3581849-3581849 on Build GRCh38
rs925478193 Chr.16: 3581868-3581868 on Build GRCh38
rs951178498 Chr.16: 3581781-3581781 on Build GRCh38
rs749668095 Chr.16: 3582031-3582031 on Build GRCh38
rs997317901 Chr.16: 3581717-3581717 on Build GRCh38
rs193089761 Chr.16: 3581809-3581809 on Build GRCh38
rs1041036368 Chr.16: 3582047-3582049 on Build GRCh38
rs942683559 Chr.16: 3582047-3582047 on Build GRCh38
rs114600718 Chr.16: 3581768-3581768 on Build GRCh38
rs868142294 Chr.16: 3581765-3581765 on Build GRCh38
rs1048091964 Chr.16: 3582067-3582067 on Build GRCh38
rs1004047755 Chr.16: 3582002-3582002 on Build GRCh38
rs1037421761 Chr.16: 3582093-3582093 on Build GRCh38
rs978734523 Chr.16: 3581979-3581979 on Build GRCh38
rs1026796213 Chr.16: 3581778-3581778 on Build GRCh38
rs567515998 Chr.16: 3582113-3582113 on Build GRCh38
rs757391667 Chr.16: 3581735-3581735 on Build GRCh38
rs868278470 Chr.16: 3581873-3581873 on Build GRCh38
rs564926554 Chr.16: 3581736-3581736 on Build GRCh38
rs950467431 Chr.16: 3581721-3581721 on Build GRCh38
rs997771598 Chr.16: 3581864-3581864 on Build GRCh38

View Full Product Details