The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000579.3 | NP_000570.1 | 2 - 3 | 345 | 98 |
NM_001100168.1 | NP_001093638.1 | 2 - 3 | 110 | 98 | |
GenBank mRNA | AK292132.1 | - | 1 - 2 | 42 | 98 |
AK314882.1 | - | 1 - 2 | 134 | 98 | |
BC038398.1 | - | 1 - 2 | 31 | 98 | |
DA818906.1 | - | 1 - 2 | 134 | 98 | |
U54994.1 | - | 1 - 2 | 42 | 98 |