The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001306074.1 | NP_001293003.1 | 4 - 5 | 933 | 88 |
NM_001306075.1 | NP_001293004.1 | 4 - 5 | 921 | 88 | |
NM_006527.3 | NP_006518.1 | 5 - 6 | 1038 | 88 | |
GenBank mRNA | AK091735.1 | - | 4 - 5 | 498 | 88 |
AK093314.1 | - | 5 - 6 | 511 | 88 | |
AK094257.1 | - | 5 - 6 | 852 | 88 | |
AK300826.1 | - | 4 - 5 | 511 | 88 | |
AK315635.1 | - | 5 - 6 | 609 | 88 | |
BC014908.1 | - | 5 - 6 | 607 | 88 | |
BC015703.1 | - | 5 - 6 | 607 | 88 | |
BT007433.1 | - | 5 - 6 | 485 | 88 | |
DB031118.1 | - | 5 - 6 | 511 | 88 | |
U75679.1 | - | 5 - 6 | 600 | 88 | |
Z71188.1 | - | 5 - 6 | 589 | 88 |