The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001086.2 | NP_001077.2 | 2 - 3 | 447 | 123 |
XM_005247104.4 | XP_005247161.1 | 2 - 3 | 461 | 123 | |
GenBank mRNA | AK290628.1 | - | 2 - 3 | 398 | 123 |
AV705031.1 | - | 2 - 3 | 447 | 123 | |
BC032309.1 | - | 2 - 3 | 382 | 123 | |
L32179.1 | - | 2 - 3 | 442 | 123 |