Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AQP8
Entrez Gene ID
343
Gene Name
aquaporin 8
Gene Aliases
AQP-8
Chromosome Location
Chr.16: 25216917 - 25228932 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001169.2 NP_001160.2 2 - 3 339 73
XM_011545822.2 XP_011544124.1 2 - 3 382 73
GenBank mRNA AB013456.1 - 2 - 3 341 73
AF067797.1 - 2 - 3 342 73
AK222964.1 - 2 - 3 304 73
AK222975.1 - 2 - 3 336 73
BC040630.1 - 2 - 3 307 73

View Full Product Details