The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001161705.1 | NP_001155177.1 | 6 - 7 | 857 | 88 |
NM_001195.4 | NP_001186.1 | 6 - 7 | 1001 | 88 | |
NM_001278606.1 | NP_001265535.1 | 7 - 8 | 992 | 88 | |
NM_001278607.1 | NP_001265536.1 | 6 - 7 | 853 | 88 | |
NM_001278608.1 | NP_001265537.1 | 8 - 9 | 978 | 88 | |
XM_017028005.1 | XP_016883494.1 | 5 - 6 | 916 | 88 | |
GenBank mRNA | AF039655.1 | - | 6 - 7 | 961 | 88 |
AK304673.1 | - | 8 - 9 | 978 | 88 | |
AK316247.1 | - | 7 - 8 | 870 | 88 | |
BC041483.1 | - | 6 - 7 | 857 | 88 | |
BQ230198.1 | - | 6 - 7 | 696 | 88 | |
Y16717.2 | - | 6 - 7 | 961 | 88 |