Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EREG
Entrez Gene ID
2069
Gene Name
epiregulin
Gene Aliases
EPR, ER, Ep
Chromosome Location
Chr.4: 74365143 - 74388760 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001432.2 NP_001423.1 1 - 2 232 118
GenBank mRNA AK314959.1 - 1 - 2 232 118
BC136404.1 - 1 - 2 189 118
BC136405.1 - 1 - 2 189 118
CR541887.1 - 1 - 2 66 118
D30783.1 - 1 - 2 232 118

View Full Product Details