The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001609.3 | NP_001600.1 | 1 - 2 | 163 | 69 |
XM_017016194.1 | XP_016871683.1 | 1 - 2 | 105 | 69 | |
GenBank mRNA | AK314241.1 | - | 1 - 2 | 59 | 69 |
KU177879.1 | - | 1 - 2 | 46 | 69 | |
U12778.1 | - | 1 - 2 | 61 | 69 |