The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001655.4 | NP_001646.2 | 1 - 2 | 170 | 132 |
XM_005271542.3 | XP_005271599.1 | 1 - 2 | 194 | 132 | |
GenBank mRNA | AK312298.1 | - | 1 - 2 | 148 | 132 |
BC093636.1 | - | 1 - 2 | 97 | 132 | |
BC093638.1 | - | 1 - 2 | 94 | 132 | |
BI560739.1 | - | 1 - 2 | 166 | 132 | |
DB216489.1 | - | 1 - 2 | 170 | 132 | |
X81197.1 | - | 1 - 2 | 126 | 131 |