The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001673.4 | NP_001664.3 | 1 - 2 | 195 | 100 |
GenBank mRNA | BC014621.2 | - | 1 - 2 | 71 | 100 |
BG718826.1 | - | 1 - 2 | 165 | 100 | |
DC297828.1 | - | 1 - 2 | 195 | 100 | |
M27396.1 | - | 1 - 2 | 122 | 100 |