The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001709.4 | NP_001700.2 | 1 - 2 | 323 | 143 |
GenBank mRNA | AY054398.2 | - | 1 - 2 | 264 | 143 |
EF689015.1 | - | 1 - 2 | 323 | 143 | |
X60201.1 | - | 1 - 2 | 273 | 143 | |
X91251.1 | - | 1 - 2 | 261 | 142 |