The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001853.3 | NP_001844.3 | 5 - 6 | 311 | 110 |
XM_011528545.1 | XP_011526847.1 | 5 - 6 | 309 | 110 | |
XM_017027666.1 | XP_016883155.1 | 5 - 6 | 309 | 110 | |
GenBank mRNA | BC011705.2 | - | 5 - 6 | 311 | 110 |
L41162.1 | - | 5 - 6 | 308 | 110 |