Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CTRB1
Entrez Gene ID
1504
Gene Name
chymotrypsinogen B1
Gene Aliases
CTRB
Chromosome Location
Chr.16: 75218986 - 75224924 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC005385.1 - 1 - 2 94 122
M24400.1 - 1 - 1 68 122

Gene Symbol
CTRB2
Entrez Gene ID
440387
Gene Name
chymotrypsinogen B2
Gene Aliases
-
Chromosome Location
Chr.16: 75204096 - 75207174 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025200.3 NP_001020371.3 1 - 2 85 122
GenBank mRNA AK225933.1 - 1 - 2 72 122
AK291822.1 - 1 - 2 70 122
BP324399.1 - 1 - 2 85 122

View Full Product Details