The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001039350.2 | NP_001034439.1 | 16 - 17 | 2062 | 78 |
NM_001290252.1 | NP_001277181.1 | 14 - 15 | 1629 | 78 | |
NM_001936.4 | NP_001927.3 | 16 - 17 | 1764 | 78 | |
NM_130797.3 | NP_570629.2 | 16 - 17 | 2070 | 78 | |
XM_017011808.1 | XP_016867297.1 | 17 - 18 | 2016 | 78 | |
XM_017011809.1 | XP_016867298.1 | 17 - 18 | 1779 | 78 | |
XM_017011810.1 | XP_016867299.1 | 17 - 18 | 1882 | 78 | |
XM_017011811.1 | XP_016867300.1 | 17 - 18 | 1806 | 78 | |
XM_017011812.1 | XP_016867301.1 | 12 - 13 | 1177 | 78 | |
GenBank mRNA | AK293582.1 | - | 9 - 10 | 941 | 78 |
AK294483.1 | - | 14 - 15 | 1399 | 78 | |
BC150304.1 | - | 16 - 17 | 2061 | 78 | |
BX647966.1 | - | 16 - 17 | 1827 | 78 | |
DA911296.1 | - | 4 - 5 | 323 | 78 | |
M96859.1 | - | 16 - 17 | 1811 | 78 | |
M96860.1 | - | 16 - 17 | 1658 | 78 |