Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HK3
Entrez Gene ID
3101
Gene Name
hexokinase 3
Gene Aliases
HKIII, HXK3
Chromosome Location
Chr.5: 176880869 - 176899373 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002115.2 NP_002106.2 1 - 2 68 73
XM_011534540.2 XP_011532842.1 1 - 2 109 73
GenBank mRNA BC028129.1 - 1 - 2 68 73
U51333.1 - 1 - 2 50 73

View Full Product Details