Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HLA-DPB1
Entrez Gene ID
3115
Gene Name
major histocompatibility complex, class II, DP beta 1
Gene Aliases
DPB1, HLA-DP, HLA-DP1B, HLA-DPB
Chromosome Location
Chr.6: 33075926 - 33089696 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002121.5 NP_002112.3 1 - 2 217 105
GenBank mRNA AB208978.1 - 1 - 2 133 105
AK291986.1 - 1 - 2 105
AK301322.1 - 1 - 2 178 105
AK303153.1 - 1 - 2 105
AK313305.1 - 1 - 2 155 105
AY656678.1 - 1 - 2 101 105
AY656679.1 - 1 - 2 101 105
AY656680.1 - 1 - 2 105
AY804132.1 - 1 - 2 148 105
AY804133.1 - 1 - 2 105
AY804134.1 - 1 - 2 138 105
AY804135.1 - 1 - 2 105
AY804136.1 - 1 - 2 104 105
AY804137.1 - 1 - 2 138 105
AY804138.1 - 1 - 2 138 105
AY804139.1 - 1 - 2 105
AY804140.1 - 1 - 2 105
AY804141.1 - 1 - 2 105
AY804142.1 - 1 - 2 148 105
AY804143.1 - 1 - 2 105
AY831402.1 - 1 - 2 149 105
AY831403.1 - 1 - 2 105
AY831404.1 - 1 - 2 101 105
BC007963.2 - 1 - 2 134 105
BC013184.1 - 1 - 2 105
BC015000.1 - 1 - 2 105
BP213121.1 - 1 - 2 217 105
BT019780.1 - 1 - 2 105
DQ284441.1 - 1 - 2 136 105
FR775797.1 - 1 - 2 105
GQ891549.1 - 1 - 2 105
HE603611.1 - 1 - 2 105
HE603612.1 - 1 - 2 101 105
HE614584.1 - 1 - 2 101 105
HE614585.1 - 1 - 2 105
HE614586.1 - 1 - 2 101 105
HE614587.1 - 1 - 2 105
HE614588.1 - 1 - 2 105
HE614589.1 - 1 - 2 105
HE614590.1 - 1 - 2 105
HE614591.1 - 1 - 2 105
J03041.1 - 1 - 2 170 105
M28200.1 - 1 - 2 105
M28202.1 - 1 - 2 77 105
M83664.1 - 1 - 2 105
X01426.1 - 1 - 2 170 105
X03067.1 - 1 - 2 149 105

View Full Product Details