The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002299.3 | NP_002290.2 | 2 - 3 | 736 | 79 |
XM_017004088.1 | XP_016859577.1 | 2 - 3 | 756 | 79 | |
GenBank mRNA | X07994.1 | - | 2 - 3 | 736 | 79 |